RapidFinder™ Water Buffalo ID Kit
Thermo Scientific™

RapidFinder™ Water Buffalo ID Kit

The Thermo Scientific™ RapidFinder™ Water Buffalo ID Kit enables the simple and reliable detection of water buffalo (Bubalus bubalis) DNA in food and feed samples.

Have Questions?
Catalog NumberQuantity
A2441448 reactions
Catalog number A24414
Price (INR)
-
Quantity:
48 reactions
The RapidFinder™ Water Buffalo ID Kit enables the simple and reliable detection of Water Buffalo (Bubalus bubalis) DNA in food and feed samples.

The RapidFinder™ Water Buffalo ID Kit enables the simple and reliable detection of Water Buffalo DNA in food and feed samples. The method works by detection of water buffalo (Bubalus bubalis) DNA that is present in food and feed samples by real-time PCR. Unlike ELISA based methods, when used with the GMO Extraction Kit (Cat. No. 4466336) for sample preparation, the RapidFinder™ Water Buffalo ID Kit detects and provides sensitivity down to 0.01% of water buffalo (Bubalus bubalis) DNA. The included positive control in the kit, which comprises 0.1% Water Buffalo DNA, can be used as a reference to set the threshold of detection of the assay.

  • Simple- Real-time PCR based assay, no electrophoresis needed
  • Flexible- Water Buffalo DNA detected in any food or feed sample, both raw and processed
  • Reliable- Internal Positive Control (IPC) provides process control check for PCR inhibitors
  • Fast- From sample to result in the same day

The assay is compatible with Applied Biosystems™ QuantStudio 5™ and 7500 Fast Real-Time PCR Systems.

Sample Preparation

DNA sample preparation is accomplished with the easy-to-use GMO Extraction Kit (Cat. No. 4466336). Using up to 20 g of sample rather than the smaller sample sizes accommodated by most other DNA isolation procedures minimizes sampling errors. The GMO Extraction Kit is compatible with raw and processed meat, as well as composite food samples.

Part of a Complete Portfolio of Solutions for Animal Species Detection

The RapidFinder™ Water Buffalo ID Kit is just one of our many available kits for the sensitive detection of various animal species in food and feed samples. Other RapidFinder™ animal species identification kits available include those for the detection of pork, beef, horse, goat, rabbit, fallow deer, duck, sheep, ruminant, chicken, poultry, and turkey.

The RapidFinder™ Species Identification Kits are made available through a partnership with Health in Code, S.L.

For Testing of Food and Environmental Samples Only.
Specifications
DescriptionWater Buffalo ID Kit
For Use With (Equipment)QuantStudio 5™ and 7500 Fast
Product LineRapidFinder
Product TypeRapidFinder™
Quantity48 reactions
Sample TypeFood and Environmental Samples
Sensitivity0.01% of Water Buffalo (Bubalus bubalis) DNA
Shelf Life2 years
SpeciesBubalus bubalis
Detection MethodPrimer-probe
For Use With (Application)Food and Beverage
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Contains 1 tube each of: Water Buffalo Master Mix (store at -20°C) General Master Mix (store at -20°C upon receipt; 2-8 °C after initial use) Positive Control (store at -20°C)