RapidFinder™ Tuna ID Kit
Thermo Scientific™

RapidFinder™ Tuna ID Kit

The Thermo Scientific™ RapidFinder™ Tuna ID Kit enables the simple and reliable detection of tuna (genus Thunnus) DNA in food and feed samples.

Have Questions?
Catalog NumberQuantity
A2441548 reactions
Catalog number A24415
Price (INR)
-
Quantity:
48 reactions
Request bulk or custom format
Specifications
DescriptionTuna ID Kit
For Use With (Equipment)QuantStudio 5™ and 7500 Fast
Product LineRapidFinder
Quantity48 reactions
Sample TypeFood and Environmental Samples
View more
The RapidFinder™ Tuna ID Kit enables the simple and reliable detection of tuna DNA (genus Thunnus) in food and feed samples. The kit detects samples containing ≥0.01% of tuna DNA.

The RapidFinder™ Tuna ID Kit enables the simple and reliable detection of tuna DNA in food and feed samples. The method works by detection of tuna (genus Thunnus) DNA that is present in food and feed samples by real-time PCR. Unlike ELISA based methods, when used with the GMO Extraction Kit (Cat. No. 4466336) for sample preparation, the RapidFinder™ Tuna ID Kit detects blends containing ≥1% of tuna DNA.

  • Simple-real-time PCR based assay, no electrophoresis needed
  • Flexible-tuna DNA detected in any food or feed sample, both raw and processed
  • Reliable-Internal Positive Control (IPC) provides process control check for PCR inhibitors
  • Fast-from sample to result in the same day

The assay is compatible with Applied Biosystems™ QuantStudio 5™ and 7500 Fast Food Safety Real-Time PCR systems.

Sample Preparation

DNA sample preparation is accomplished with the easy-to-use GMO Extraction Kit (Cat. No. 4466336). Using up to 20 g of sample rather than the smaller sample sizes accommodated by most other DNA isolation procedures minimizes sampling errors. The GMO Extraction Kit is compatible with raw and processed meat, as well as composite food samples.

Part of a Complete Portfolio of Solutions for Fish Species Detection

The RapidFinder™ Pollack ID Kit is just one of our many available kits for the sensitive detection of various fish species in food and feed samples. Other RapidFinder™ Fish species identification kits available include those for the detection of Basa, Gadus, Haddock, Pollock, Pollack, and Specific Tuna.

For Testing of Food and Environmental Samples Only.
Contents & Storage

Contains 1 tube each of: Tuna Master Mix (store at -20°C) General Master Mix (store at -20°C upon receipt; 2-8 °C after initial use) Positive Control (store at -20°C)