QuantStudio™ 5 10-Dye Spectral Calibration Kit, 384-well
Applied Biosystems™

QuantStudio™ 5 10-Dye Spectral Calibration Kit, 384-well

This easy-to-use dye calibration kit establishes the pure dye spectra and multicomponent values for FAM™, VIC™, ROX™, TAMRA™, SYBR™ ,Read more
Have Questions?
Catalog NumberQuantity
A263411 kit
Catalog number A26341
Price (INR)
114,889.00
Each
Quantity:
1 kit
Price (INR)
114,889.00
Each
This easy-to-use dye calibration kit establishes the pure dye spectra and multicomponent values for FAM™, VIC™, ROX™, TAMRA™, SYBR™ , ABY™, JUN™, MUSTANG PURPLE™, NED™, and Cy™5 dyes on the Applied Biosystems™ QuantStudio™ 5 384-well real-time PCR system.

The QuantStudio™ 5 10-Dye Spectral Calibration Kit contains the following:

• QuantStudio™ 5 Spectral Calibration Plate 1 (FAM™, VIC™, ROX™, TAMRA™, and SYBR™ dyes), 384-well (Cat. No. A26334)
• QuantStudio™ 5 Spectral Calibration Plate 2 (ABY™, JUN™, MUSTANG PURPLE™, NED™, and Cy™5 dyes), 384-well (Cat. No. A26335)

This multiple-dye plate calibration kit enables simultaneous dye calibrations in single plates, significantly reducing the effort and time required for dye calibration. In addition, the formulations of the dyes in Plate 2 improve results with multiplexing by more accurately representing the fluorescent spectra of the respective probes found in your real-time PCR experiments.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Calibrated DyeCy™5, FAM, NED, ROX, TAMRA™, VIC, SYBR™ Green
Calibration TypeSpectral Calibration
For Use With (Application)Spectral Calibration
For Use With (Equipment)QuantStudio™ 5, QuantStudio™ 6 Pro, QuantStudio™ 7 Pro, QuantStudio™ 6 Flex (Software version 1.6 or later), QuantStudio™ 7 Flex (Software version 1.6 or later)
Format384-well
GC-Rich PCR PerformanceLow
PCR MethodqPCR
Plate Compatibility384-well
Quantity1 kit
Shipping ConditionDry Ice
Product TypeCalibration Plate
Unit SizeEach
Contents & Storage
Store at –20°C ± 5°C.