Avidin, NeutrAvidin™, FITC conjugate
Invitrogen™

Avidin, NeutrAvidin™, FITC conjugate

Searching for superior alternatives to fluorescein? Our Alexa Fluor Dye Series offers everything you're looking for and more.Read more
Have Questions?
Catalog NumberQuantity
A2662
also known as A-2662
1 mg
Catalog number A2662
also known as A-2662
Price (INR)
17,134.00
線上優惠
Ends: 31-Dec-2026
28,556.00
Save 11,422.00 (40%)
Each
Quantity:
1 mg
Price (INR)
17,134.00
線上優惠
Ends: 31-Dec-2026
28,556.00
Save 11,422.00 (40%)
Each
Searching for superior alternatives to fluorescein? Our Alexa Fluor Dye Series offers everything you're looking for and more.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 mg
Shipping ConditionRoom Temperature
ConjugateFITC (Fluorescein)
Product LineNeutrAvidin
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (13)

Citations & References
Abstract
Flow-cytometric isolation of human antibodies from a nonimmune Saccharomyces cerevisiae surface display library.
Authors:Feldhaus MJ, Siegel RW, Opresko LK, Coleman JR, Feldhaus JM, Yeung YA, Cochran JR, Heinzelman P, Colby D, Swers J, Graff C, Wiley HS, Wittrup KD
Journal:Nat Biotechnol
PubMed ID:12536217
'A nonimmune library of 10(9) human antibody scFv fragments has been cloned and expressed on the surface of yeast, and nanomolar-affinity scFvs routinely obtained by magnetic bead screening and flow-cytometric sorting. The yeast library can be amplified 10(10)-fold without measurable loss of clonal diversity, allowing its effectively indefinite expansion. The ... More
Randomly ordered addressable high-density optical sensor arrays.
Authors:Michael KL, Taylor LC, Schultz SL, Walt DR
Journal:Anal Chem
PubMed ID:9553489
'Array-based sensors provide an architecture for multianalyte sensing. In this paper, we report a new approach for array fabrication. Sensors are made by immobilizing different reactive chemistries on the surfaces of microspheres. Sensor arrays are prepared by randomly distributing a mixture of microsphere sensors on an optical substrate containing thousands ... More
Interphase cytogenetics in pathology: principles, methods, and applications of fluorescence in situ hybridization (FISH).
Authors:Werner M, Wilkens L, Aubele M, Nolte M, Zitzelsberger H, Komminoth P
Journal:Histochem Cell Biol
PubMed ID:9387931
'Characteristic chromosome aberrations have been identified in various tumors. Fluorescence in situ hybridization (FISH) using specific probes that are generated by vector cloning or in vitro amplification and labeled with fluorescent dyes allow for the detection of these genetic changes in interphase cells. This technique, that is also referred to ... More
Improved immunocytochemical detection of biotinylated probes with Neutralite avidin.
Authors:van Roy N, Mangelschots K, Speleman F
Journal:Trends Genet
PubMed ID:8488564
Adhesive and mammalian transglutaminase substrate properties of Candida albicans Hwp1.
Authors:Staab JF, Bradway SD, Fidel PL, Sundstrom P
Journal:Science
PubMed ID:10066176
The pathogenesis of candidiasis involves invasion of host tissues by filamentous forms of the opportunistic yeast Candida albicans. Morphology-specific gene products may confer proinvasive properties. A hypha-specific surface protein, Hwp1, with similarities to mammalian small proline-rich proteins was shown to serve as a substrate for mammalian transglutaminases. Candida albicans strains ... More