3500 Dx Genetic Analyzer
Applied Biosystems™
3500 Dx Genetic Analyzer
The 3500 Dx Genetic Analyzer is an 8-capillary CE-IVD-marked member of the Applied Biosystems™ 3500 series of genetic analyzers basedRead more
Have Questions?
Catalog NumberQuantity
A277721 System
Catalog number A27772
Price (INR)
-
Quantity:
1 System
The 3500 Dx Genetic Analyzer is an 8-capillary CE-IVD-marked member of the Applied Biosystems™ 3500 series of genetic analyzers based on capillary electrophoresis.
Reliability
• High accuracy—using BigDye™ Terminator v1.1 or v3.1 Sequencing Standards, the instrument has shown >99% read accuracy
• Proven results—Applied Biosystems genetic analyzers are a trusted standard for both Sanger sequencing and fragment analysis
Flexibility
• Provides a dual mode user interface: the Diagnostic Mode and the Research Use Only Mode
• Enables both fragment analysis and Sanger sequencing in the secure Diagnostic Mode
• Enables import of validated sequencing and fragment analysis assay protocols to run in Diagnostic Mode
• Offers different configurations to meet your throughput needs: this 8-capillary version and a 24-capillary version
Simplicity and efficiency
• Reduced instrument hands-on time with simplified installation
• Controlled and efficient run setup with intuitive user interface, preconfigured plate templates, and software-controlled functions (plate setup, data collection, analysis)
• Maintenance calendar with programmable prompts and service history functionality
• RFID (Radio Frequency Identification) tracking of consumables, built-in quality controls, and real-time data quality evaluation
• SAE (Security, Audit, and Electronic) signature capability to assist with compliance to governing regulations
The 3500 Dx Genetic Analyzer is a trusted capillary electrophoresis-based platform for development of your assays. Capillary electrophoresis-based applications are supported by a large number of publications over the many years that they have been used successfully in laboratory testing. The 3500 Dx Genetic Analyzer is a partner in your laboratory, making it easy for you to comply with various industry standards and train new users.
The 3500 Dx Genetic Analyzer includes a computer system and associated data collection software, as well as an installation kit with the initial set of accessories and reagents needed to start using the instrument right away. It is backed by a comprehensive service and support program with options to purchase additional services per your laboratory maintenance and audit requirements.
Intended use
The Applied Biosystems 3500 Dx/3500xL Dx Genetic Analyzers are intended for use by trained operators in the analysis of human DNA or RNA, for the detection of genetic changes that may lead to disease presence or susceptibility.
Note: The 3500 Dx Genetic Analyzer data collection software uses the Windows™ 7 operating system.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)3500 Dx Genetic Analyzer
Product TypeGenetic Analyzer
Quantity1 System
Shipping ConditionRoom Temperature
SpeciesHuman
Unit SizeEach
Contents & Storage
This system package consists of an 8-capillary instrument, Data Collection Software, and a Dell Workstation and monitor.
Frequently asked questions (FAQs)
When I started my run on the Applied Biosystems 3500/3500xL Genetic Analyzer, an error occurred: "Unstable Electrophoresis current detected, check for air bubbles" but I don't see any bubbles. What else can cause this?
Some of the other causes of an Unstable Electrophoresis current detected error message are:
1.Leak on the system
2. Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3. Running Buffer that was:
-On the instrument longer than 2 weeks
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues
Inspect the system for leaks. If you do not see any leaks on the system, perform the wash the pump chamber and channels wizard using the conditioning pouch and place fresh, non-expired polymer and fresh Anode and Cathode buffer containers. If the problem persists, a service call may be required.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
Do I need to replace the buffer every 14 days if the Applied Biosystems 3500/3500xL Genetic Analyzer is not in use?
If the instrument is not in use, it is not necessary to replace the buffer every 14 days. However, the buffer level needs to remain at the fill line to prevent the capillary or array from drying out. Top off the buffer volume with water if necessary. Replace the anode and cathode buffer containers prior to starting a new run.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.