QuantStudio™ 5 Real-Time PCR System, 96-well, 0.1 mL
Applied Biosystems™

QuantStudio™ 5 Real-Time PCR System, 96-well, 0.1 mL

The Applied Biosystems QuantStudio 5 Real-Time PCR System is designed for users who need superior performance, maximum dye versatility, andRead more
Have Questions?
Catalog NumberQuantity
A281381 system
Catalog number A28138
Price (INR)
-
Quantity:
1 system
The Applied Biosystems QuantStudio 5 Real-Time PCR System is designed for users who need superior performance, maximum dye versatility, and security options in a real-time PCR system that is affordable and easy to use. The optimized Design and Analysis software is ideal for both first-time and experienced users. When connected to iConnect, Thermo Fisher's cloud-based platform, the QuantStudio 5 system provides access to your data wherever and whenever you want. Using proven OptiFlex technology (featuring six decoupled channels and white LED) and featuring six independent Veriflex temperature zones, the QuantStudio 5 96-well system enables improved data accuracy and sensitivity for a broad range of genomic applications. The QuantStudio 5 system also offers the built-in software features of electronic record security and prevention of unauthorized instrument access to assist with 21 CFR Part 11 compliance.

Quantstudio 3/5 Firmware and Design & Analysis Software >

Features of the QuantStudio 5 system with 96-well block include:

• Proven OptiFlex technology (six decoupled channels/21 filter combinations with white LED) for trusted results
• User account management, locked workflow, and Pause features for greater control of experiment data
• Built-in software features to support compliance with 21 CFR Part 11 guidelines
• Secure cloud-based software provides increased accessibility and storage options
• Simplified instrument software with interactive touchscreen for greater ease of use
• 96-well 0.2 mL format or 96-well 0.1 mL format

iConnect
iConnect provides scientists with a secure place to store, analyze, and share data. Upload data from the instrument or upload your own files from anywhere you have web access. Applied Biosystems qPCR Analysis Modules within iConnect are web browser-based software tools that are flexible, fast and easy to use.

Easy to use
The QuantStudio 5 Real-Time PCR System is compact in form, factory calibrated and is delivered in user-friendly packaging to enable fast and simple installation for immediate use. The interactive touchscreen provides a modern usability experience and the simplified instrument software/pre-optimized protocol templates allow users of any experience level to quickly set up and start a run.

Design and analysis software options:

Option 1: QuantStudio Design and Analysis cloud software
• Supports web browser-based system configuration with PC or MAC computer
• Streamlined software for improved usability and response time
• Enables secure access of your data when and where you want through the cloud
• No software to install, no licenses and no versions to update

Option 2: QuantStudio Design and Analysis desktop software
• Supports traditional co-located computer system configuration on Windows 10 OS
• Streamlined software for improved usability and response time
• No licenses to purchase

Total control
With built-in software features to support compliance with 21 CFR Part 11 guidelines, user account management, locked workflow and Pause features, the QuantStudio 5 system offers users greater control of experiment data

Multiplex performance results that you can trust
Improved well-to-well and instrument-to-instrument data accuracy is achieved with OptiFlex technology. In the 96-well system this technology features six decoupled excitation and emission filter channels, for a total of 21 filter combinations, with white LED.

Sensitive data analysis
Detect changes in target quantity as small as 1.5-fold in single-plex reactions and obtain 10 logs of linear dynamic range.

High application versatility
Simple and optimized protocols, reliable assays and reagents, and intuitive software are available for a broad range of applications, including gene expression, genetic variation, gene regulation, and more.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Display TypeTouchscreen
For Use With (Equipment)QuantStudio™
Format96-well Plate
Product LineQuantStudio
Quantity1 system
Shipping ConditionRoom Temperature
Volume (Metric) Thermal Block Sample0.1 ml
Block FormatNon-interchangeable
Unit SizeEach

Frequently asked questions (FAQs)

What is the difference between continuous melt curves and step and hold melt curves on the Applied Biosystems Real-Time PCR Instruments?

Continuous melt curve: increases the temperature by the ramp increment (degrees C/sec) so that the temperature will change at a constant rate set in the protocol.
Step and Hold melt curve: increases the temperature by the ramp increment (degrees C) and then holds at that temperature for the time specified by the user.

The amount of data collected will depend on the filter(s) selected. The more filters you use, the longer the acquisition will take and the fewer data points will be collected.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

The calibration on my real-time PCR instrument has expired and I get an error message to calibrate my machine. Is it possible to turn off the reminders and carry on with my experiments without calibrating the machine?

Yes, you can click okay to the message and continue with the run and calibrate the instrument at a later time. The exception is that the QuantStudio Dx, QuantStudio 5 Dx, and QuantStudio 7 Pro Dx instruments cannot be run in IVD mode with expired calibrations.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center

What is the admin pin for QuantStudio 3 and 5 Real-Time PCR Systems?

If the admin PIN has not been changed, 1234 should work. If it does not work, you can also try 0000.
However, if the admin PIN has been changed and none of the above PINs work, follow the instructions in the QuantStudio 3 and 5 Real-Time PCR Systems manual, page 100.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

How do I download the Pathogen Interpretive Software?

Once you place your order, please contact our Instrument Service team at 1-800-955-6288 option 3, 1 (this kit is only available in North America). You will be given an access code to the TaqPath Monkeypox/Orthopox Virus DNA Kit eLearning where you’ll learn how to install, update, and download the Pathogen Interpretive Software package for the TaqPath Monkeypox/Orthopox Virus DNA Kit.

With the TaqPath Monkeypox/Orthopox Virus DNA Kit, what software should I use for data analysis?

Results are analyzed using the following software:
- Applied Biosystems Pathogen Interpretive Software v1.1
- SAE Administrator Console Dx v1.0 or v1.2 (for security and audit functions)
- Pathogen Interpretive Software 1.0.0 DAT (SAE Profile)

Please use the appropriate assay panel and template for your instrument:
- MPXOPX-EUA_QS5-9601_1.0.0.zip
- MPXOPX-EUA_QS5Dx-9602_1.0.0.zip
- MPXOPX-EUA_QS7F-384_1.0.0.zip

Note: These zip files are located in EducationConnect (www.thermofisher.com/educationconnect). The eLearning course needs to be completed to be granted access.