The Applied Biosystems™ TaqMan™ RNase P Assay with VIC™ dye-labeled QSY™ probe provides a pre-formulated assay for quantitating human RNaseRead more
Have Questions?
Catalog Number
Quantity
A30064
250 reactions
Catalog number A30064
Price (INR)
42,636.00
Each
Quantity:
250 reactions
Price (INR)
42,636.00
Each
The Applied Biosystems™ TaqMan™ RNase P Assay with VIC™ dye-labeled QSY™ probe provides a pre-formulated assay for quantitating human RNase P. This assay enables relative gene expression quantification in cDNA samples when used with other gene expression assays. It consists of a VIC dye-labeled QSY probe plus sequence-specific forward and reverse primers. The assay can be used for multiplex or singleplex PCR reactions. For an assay with ABY™ dye-labeled probe rather than VIC dye-labeled probe, see Cat. No. 4485714.
Benefits of this assay include:
• Eliminate assay design time using pre-designed primers and pre-designed VIC dye-labeled TaqMan probe • Minimize experimental optimization time • Use in conjunction with FAM™ dye-labeled predesigned gene expression assays
Save time in assay design, formulation, and testing by using the TaqMan RNase P Assay as your control when quantifying gene expression. This pre-designed probe and primer set enables you to normalize the amount of sample RNA or DNA in a reaction when used with a control sample. Or use it for relative gene expression quantification in cDNA samples when used with other gene expression assays.
The VIC dye is optimized for use with the second filter of our QuantStudio™, ViiA™ 7, and 7500 series real-time PCR instruments.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
No. of Reactions250 Reactions
PrimerReal-Time PCR Primers
Product LineQSY, TaqMan, VIC
Product TypeqPCR Master Mix
Quantity250 reactions
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice