Product Image
Invitrogen™

Silencer™ Human Tumor Suppressor siRNA Library

Mutations that cause a loss or reduction in gene function can result in the progression to cancer, usually in combination with other genetic changes. Tumor suppressor genes, or anti-oncogenes, can protect cells on the path to becoming cancerous.
Have Questions?
Catalog NumberQuantity
A301471 Set
Catalog number A30147
Price (INR)
1,177,044.00
Online offer
Ends: 31-Dec-2026
1,961,739.80
Save 784,695.80 (40%)
Each
Quantity:
1 Set
Price (INR)
1,177,044.00
Online offer
Ends: 31-Dec-2026
1,961,739.80
Save 784,695.80 (40%)
Each

Mutations that cause a loss or reduction in gene function can result in the progression to cancer, usually in combination with other genetic changes. Tumor suppressor genes, or anti-oncogenes, can protect cells on the path to becoming cancerous. The Silencer™ Human Tumor Suppressor siRNA Library contains 2,175 Ambion™ Silencer siRNAs targeting 725 tumor suppressor genes. The library was designed using the most up-to-date genome databases, including the NCBI RefSeq database and cross-referenced to the Gene Ontology Consortium (GO) database and/or the HUGO Gene Nomenclature Committee (HGNC) as well as the Tumor Suppressor Gene Database (TSGene). Utilizing Ambion Silencer technology built on stringent bioinformatics to enable high target specificity and siRNA potency, the Silencer Human Tumor Suppressor siRNA Library is an ideal solution for focused and cost-effective RNAi screening against tumor suppressor genes.

  • Comprehensive – Expedite your tumor suppressor RNAi functional screens with pre-defined Silencer Human Tumor Suppressor siRNA Library
  • Reliable – Increase target screening accuracy with three individual siRNAs per gene target
  • Effective – Reduce off target impacts at low concentrations with proven Ambion Silencer siRNA design
  • Convenient – Reduce data de-convolution and accelerate target validation with our arrayed format
  • Customizable – Choose the best option for your workflow demands, including additional siRNA scales and pooled formats

Why use the Silencer Human Tumor Suppressor siRNA Library?
Together with careful bioinformatics selection of gene targets, Silencer siRNAs are optimized with design and high quality synthesis with guaranteed silencing. The Silencer Human Tumor Suppressor siRNA Library contains our pre-designed and when available, validated siRNAs generated with maximum potency and specificity in mind so you can focus on data quality. All Silencer libraries are selected against the latest available databases and resources for up-to-date content. Full siRNA sequence information, as well as the exon targeted and each targeted gene's symbol, full name, aliases, RefSeq number, and Locus ID, plus results from any validation experiments performed using the included siRNAs is provided. Please contact us for the complete list of targets.

Silencer siRNA libraries allow you to screen efficiently and maximize data output
Ambion Silencer siRNAs are designed with potency in mind to help ensure gene knockdown at minimum concentrations. This not only helps ensure less potential for off target effects and false positive data but also minimizes false negative outcomes of siRNA screening. The Silencer Human Tumor Suppressor siRNA Library contains 0.25 nmol of each siRNA, sufficient for 100 transfections (at 25 nM siRNA, 100 μL transfection volume).

Flexibility to meet your workflow needs
Thermo Fisher Scientific offers a comprehensive list of pre-defined or custom siRNA libraries to human, mouse or rat genes with 1, 2, or 5 nmol of each siRNA or larger sizes. For a complete gene list, additional information or to discuss your research with one of our technical experts, contact your local sales representative or e-mail us at RNAilibraries@lifetech.

Specifications
For Use With (Application)RNAi
Product LineSilencer
Product TypesiRNA Library
PurityStandard
Quantity1 Set
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Format96-well Plate
RNAi TypesiRNA
Sample TypeRNA
SpeciesHuman
Unit SizeEach
Contents & Storage
2175 total siRNAs targeting 725 genes plated onto 27 x 96-well plates

Store at or below -20°C in a non-frost-free freezer.