Bacto™ Yeast Extract 2
Gibco™
Bacto™ Yeast Extract 2
Bacto Yeast Extract 2 is a concentrate of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells. Derived from primary grownRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A30801-0350 kg
A30801-01500 g
A30801-0210 kg
Catalog number A30801-03
Price (INR)
-
Quantity:
50 kg
Bacto Yeast Extract 2 is a concentrate of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells. Derived from primary grown baker's yeast, this animal-origin free product is suitable for use in microbial fermentation. Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation.
Bacto Yeast Extract 2 is one of the most complete and versatile fermentation supplements available.
Related yeast extract products
Gibco Yeast Extract—developed to provide a product for the biotechnology/pharmaceutical market with acceptable clarity and growth-promoting characteristics. Media formulations containing yeast extract are specified in standard methods for various applications.
Difco Yeast Extract UF—ultra-filtered and specifically designed for mammalian cell culture applications. With its low endotoxin level and high content of naturally occurring B vitamins, it can be used effectively to reduce the need for fetal bovine serum. It has an endotoxin level of less than or equal to 500 EU/g.
Difco Yeast Extract Low-Dusting (LD)—designed to limit the amount of dust generated compared to other spray-dried yeast extracts
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Culture TypeMammalian, Microbial, and Insect
FormatDrum
Quantity50 kg
Shelf Life1095 days
ClassificationAnimal Origin-free
FormPowder
Product TypeYeast Supplement
SterilityNon-sterile
Unit Size50 kg