AmpFLSTR™ PCR Enhancer
Applied Biosystems™
AmpFLSTR™ PCR Enhancer
AmpFLSTR™ PCR Enhancer is a reagent that enhances PCR amplification when used with buccal samples collected using the Bode Buccal DNA Collector™ and processed using the GlobalFiler™ Express PCR Amplification Kit.
Have Questions?
Catalog NumberQuantity
A31023500 reactions
Catalog number A31023
Price (INR)
771,543.00
Each
Quantity:
500 reactions
Price (INR)
771,543.00
Each
AmpFLSTR™ PCR Enhancer is a reagent that enhances PCR amplification when used with buccal samples collected using the Bode Buccal DNA Collector™ and processed using the GlobalFiler™ Express PCR Amplification Kit. By optimizing primer binding efficiency, PCR Enhancer helps simplify data analysis and reduce time to result.
  • Optimized for Bode Buccal DNA Collector samples treated with Prep-n-Go™ Buffer and processed using the GlobalFiler Express PCR Amplification Kit
  • Simply add PCR Enhancer to the PCR reaction reagents during amplification setup without pretreatment or incubation steps
  • Helps improve first pass success when using the GeneMapper™ ID-X automated genotyping expert software system for data analysis
AmpFLSTR PCR Enhancer incorporates easily into the existing direct amplification workflows of forensic laboratories processing single-source samples.
Specifications
PolymeraseTaq DNA Polymerase
Product TypePCR Enhancer
Quantity500 reactions
Shipping ConditionApproved for shipment on dry ice
Product LineAmpFLSTR
Unit SizeEach
Contents & Storage
1.25 mL AmpFLSTR PCR Enhancer
Store at -5 to -30°C on receipt, 2 to 8°C after initial use.