Product Image
Applied Biosystems™
Quantifiler™ Automation Enhancer
Quantifiler Automation Enhancer, when used with the Quantifiler Trio DNA Quantification Kit on a robotic instrument, prevents bubbles from beingRead more
Have Questions?
Catalog NumberQuantity
A312871 each
Catalog number A31287
Price (INR)
-
Quantity:
1 each
Quantifiler Automation Enhancer, when used with the Quantifiler Trio DNA Quantification Kit on a robotic instrument, prevents bubbles from being introduced during automated pipetting and mixing.
The Quantifiler Trio DNA Quantification Kit is a robust kit that enables forensic laboratories to simultaneously obtain a quantitative and qualitative assessment of total human and human male DNA in a single, highly sensitive real-time PCR reaction.
To use, simply add the Quantifiler Automation Enhancer to the Quantifiler Trio DNA Quantification Kit mixture at a 1:1000 ratio. The mixture remains stable at 4°C for at least six months.
Specifications
Quantity1 each
Detection MethodqPCR
For Use With (Application)DNA Quantitation
Label or DyeUnlabeled
PCR MethodqPCR
Unit SizeEach