Dopaminergic Neuron Maturation Supplement
Gibco™
Dopaminergic Neuron Maturation Supplement
Dopaminergic Neuron Maturation Supplement is a component of the Gibco™ PSC Dopaminergic Neuron Differentiation Kit that is also offered separately.
Have Questions?
Catalog NumberQuantity
A314740110 mL
Catalog number A3147401
Price (INR)
90,352.00
Online offer
Ends: 31-Dec-2026
120,469.00
Save 30,117.00 (25%)
Each
Quantity:
10 mL
Price (INR)
90,352.00
Online offer
Ends: 31-Dec-2026
120,469.00
Save 30,117.00 (25%)
Each
Dopaminergic Neuron Maturation Supplement is a component of the Gibco™ PSC Dopaminergic Neuron Differentiation Kit that is also offered separately. It enables maturation of excess floor plate progenitor cells to midbrain dopaminergic neurons.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeStem Cells (Neural)
Culture TypeNeural Cell Culture
Quantity10 mL
Shipping ConditionDry Ice
FormLiquid
Product TypeCell Culture Supplement
SterilitySterile
pH6 to 8
Unit SizeEach
Contents & Storage
1 x 10 mL Dopaminergic Neuron Maturation Supplement
Frequently asked questions (FAQs)
Is cell density in Floor Plate Specification medium critical for Dopaminergic neuron differentiation?
Yes, cell density should be optimized to attain full confluency at day 7 in the specification medium to have an efficient differentiation.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can Dopaminergic Neuron Maturation medium be used to differentiate neural stem cells?
We have tested neural stem cells (NSCs) isolated from fetal tissue or derived from pluripotent stem cells (PSCs), and have seen that both populations can benefit from maturation medium (DMEM/F12 + Dopaminergic Neuron Maturation Supplement) to have nicely spread homogenous neurons with reduced progenitor population. Matured neurons can be further maintained in neurobasal medium supplemented with Dopaminergic Neuron Maturation Supplement.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Do you offer Dopaminergic Neuron differentiation service?
Yes. Please go here (https://www.thermofisher.com/us/en/home/life-science/stem-cell-research/stem-cell-services/stem-cell-differentiation-services.html).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.