Pharma Magnetic Stand-96
Pharma Magnetic Stand-96
Applied Biosystems™
Pharma Magnetic Stand-96
The Magnetic Stand-96 is designed for paramagnetic bead precipitation from standard 96-well, U-bottom microplates and 0.2 mL PCR plates with no additional accessories.
Have Questions?
Catalog NumberQuantity
A315431 Stand
Catalog number A31543
Price (INR)
-
Quantity:
1 Stand
The stand has a clear polyacryl base in a standard SBS 96-well microplate footprint. The use of 24 extremely strong permanent magnets enables easy and fast magnetic separation in as little as 30 seconds. It sits flat on a lab bench as well as in a microplate adapter on robotic workstations.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 Stand
Shipping ConditionRoom Temperature
Product LineKingFisher™ Flex
TypeMagnetic Stand
Unit SizeEach