TaqMan™ Vaginal Microbiota Amplification Control
Applied Biosystems™

TaqMan™ Vaginal Microbiota Amplification Control

The TaqMan™ Vaginal Microbiota Amplification Control is designed to evaluate the amplification efficacy of the TaqMan™ Vaginal Microbiota assays. ItRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A3204020 μL
A32913250 μL
Catalog number A32040
Price (INR)
40,332.00
Each
Quantity:
20 μL
Price (INR)
40,332.00
Each
The TaqMan™ Vaginal Microbiota Amplification Control is designed to evaluate the amplification efficacy of the TaqMan™ Vaginal Microbiota assays. It is a multi-target plasmid that includes amplicons for each of the qualified TaqMan Vaginal Microbiota assays, as well as control prokaryotic 16S rRNA and human RNase P RPPH1 gene sequences. This plasmid serves as a positive control for the TaqMan Vaginal Microbiota Assay collection. The microbial targets of the included amplicons are listed below.

The TaqMan Vaginal Microbiota Amplification Control can be included in vaginal microbiota profiling experiments to verify assay performance and to assist with troubleshooting.

Atopobium vaginae Prevotella bivia
Bacteroides fragilis Staphylococcus aureus
Bacterial vaginosis associated bacterium 2 (BVAB2) Streptococcus agalactiae
Chlamydia trachomatis Treponema pallidum
Enterococcus faecalis Candida albicans
Escherichia coli Candida dubliniensis
Gardnerella vaginalis Candida glabrata
Haemophilus ducreyi Candida krusei
Lactobacillus crispatus Candida lusitaniae
Lactobacillus gasseri Candida parapsilosis
Lactobacillus iners Candida tropicalis
Lactobacillus jensenii Mycoplasma genitalium
Megasphaera 1 Mycoplasma hominis
Megasphaera 2 Ureaplasma urealyticum
Mobiluncus curtisii Trichomonas vaginalis
Mobiluncus mulieris Herpes simplex 1
Neisseria gonorrhoeae Herpes simplex 2
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatTube(s)
Product LineTaqMan
Quantity20 μL
Shipping ConditionGel Packs
SpeciesNon-Human
Concentration105 Copies/μL
For Use With (Application)Pathogen Detection
FormLiquid
Label or DyeUnlabeled
Unit SizeEach
Contents & Storage
Store at -5 to -30°C.

Frequently asked questions (FAQs)

Do you have any positive controls for the vaginal microbiota assays?

The TaqMan Vaginal Microbiota Amplification Control (Cat. No. A32040) is a linearized multi-target plasmid with target sequences for each available vaginal microbiota profiling assay.

Which sample prep kit do you recommend when running vaginal microbiota profiling experiments?

We recommend the MagMAX DNA Multi-Sample Ultra Kit (Cat. No. A25597) for isolating DNA for vaginal microbiota profiling experiments.