MicroAmp™ 32-Well Clear Adhesive Film
Applied Biosystems™
MicroAmp™ 32-Well Clear Adhesive Film
The Applied Biosystems MicroAmp 32-Well Clear Adhesive Film is designed specifically for use with the MicroAmp TriFlex 3 x 32-WellRead more
Have Questions?
Catalog NumberQuantity
A32812100 Films
Catalog number A32812
Price (INR)
8,264.00
Online offer
Ends: 31-Dec-2026
13,773.00
Save 5,509.00 (40%)
Each
Quantity:
100 Films
Price (INR)
8,264.00
Online offer
Ends: 31-Dec-2026
13,773.00
Save 5,509.00 (40%)
Each
The Applied Biosystems MicroAmp 32-Well Clear Adhesive Film is designed specifically for use with the MicroAmp TriFlex 3 x 32-Well PCR Reaction Plate. The film provides an alternative to cap strips and seals 32-well samples securely to prevent PCR sample evaporation and well-to-well contamination.
Key features:
• Designed for use with the MicroAmp TriFlex 3 x 32-Well PCR Reaction Plate
• Same product design as the trusted MicroAmp Clear Adhesive Film for 96-well plates (Cat. No. 4306311), but pre-cut to fit 32-well plates
• Easy to apply and remove—simply peel off the backing and apply firmly to the 32-well plate
Specifications
For Use With (Equipment)MicroAmp TriFlex 3 x 32-Well PCR Reaction Plate
Product TypeFilm, Seal, or Applicator
Quantity100 Films
Shipping ConditionRoom Temperature
ColorClear
MaterialPolyethylene
No. of Wells32
Plate Compatibility32-well Plate
Product LineMicroAmp
Unit SizeEach