ATC Full-skirted Adaptor
Applied Biosystems™
ATC Full-skirted Adaptor
This full-skirted adaptor comes factory-installed on the 96-well Automated Thermal Cycler (ATC).
Have Questions?
Catalog NumberQuantity
A330451 adaptor
Catalog number A33045
Price (INR)
2,055.00
Each
Quantity:
1 adaptor
Price (INR)
2,055.00
Each
This full-skirted adaptor comes factory-installed on the 96-well Automated Thermal Cycler (ATC). It ensures that uniform pressure is applied by the heated lid onto the plate, and also ensures that the plate is ejected smoothly without sticking. This adaptor is only for use with 96-well fully skirted plates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)End-point PCR
For Use With (Equipment)Automated Thermal Cycler (ATC)
Quantity1 adaptor
Product TypeATC Full-skirted Plate Adaptor
Unit SizeEach