Precision ID GlobalFiler™ NGS STR Panel v2
Applied Biosystems™
Precision ID GlobalFiler™ NGS STR Panel v2
The Applied Biosystems Precision ID GlobalFiler NGS STR Panel v2 enables an innovative next-generation sequencing (NGS) approach to analysis ofRead more
Have Questions?
Catalog NumberQuantity
A3311496 reactions
Catalog number A33114
Price (INR)
474,117.00
Each
Quantity:
96 reactions
Price (INR)
474,117.00
Each
The Applied Biosystems Precision ID GlobalFiler NGS STR Panel v2 enables an innovative next-generation sequencing (NGS) approach to analysis of DNA mixtures for casework applications using massively parallel sequencing built on Ion AmpliSeq technology for superior efficiency. The panel includes 35 markers comprised of 21 CODIS STR markers, 9 additional multi-allelic STR markers, and 4 sex determination markers to aid in mixture interpretation of complex casework samples.
Using a minimal DNA input of 125 pg, this targeted forensic marker panel is optimized for use with the Precision ID DL8 Kit (Cat. No. A33212) and the Ion S5 Precision ID Chef & Sequencing kit (Cat. No. A33208). The automated Precision ID workflow enables streamlined sample extraction-to-genotypes using the Ion Chef System, Ion GeneStudio series sequencer, and Converge Software NGS Analysis Module.
Simplicity
• As little as 125 pg of input DNA
• Manual and automated Ion Chef library preparation
• Template preparation with the Ion Chef System
• Interpret profiles with the Converge NGS Analysis Module
Scalability
• Ion 520 or Ion 530 chip-enabled
• Ability to multiplex up to 32 samples for efficient sample processing
The Converge Software NGS module offers complete NGS data analysis of targets amplified with the Precision ID GlobalFiler NGS STR Panel v2. STR markers are displayed as both allele number and base sequence for each repeat structure, offering enhanced mixture resolution. The software also analyzes single nucleotide polymorphisms (SNPs) in flanking regions of the STR markers. This unique NGS STR panel combines maximum compatibility with CODIS core loci databasing standards and superior discrimination power, enabling forensic DNA labs worldwide to maximize information recovery and improve overall efficiency. The Precision ID GlobalFiler NGS STR Panel v2 is optimized for casework samples.
Compatible with the Ion S5/S5XL and Ion GeneStudio series systems only. Not for use with the Ion OneTouch2 System or Ion PGM System.
NOTE: The GlobalFiler NGS STR Panel v2 provides sufficient material to perform 96 reactions using the manual Precision ID Library kit (Cat. No. A26435) or 32 reactions using the automated Precision ID DL8 Library Kit (Cat. No. A33212).
Specifications
Product TypePrecision ID NGS STR Panel v2
Quantity96 reactions
Sequencing TypeGenome & DNA Sequencing
Shipping ConditionDry Ice
Throughput32 Samples
Product LineGlobalFiler
Starting MaterialDNA
Unit SizeEach
Contents & Storage
Contains Precision ID GlobalFiler NGS STR Panel (1 tube), store at -5 to -30°C
The GlobalFiler NGS STR Panel v2 provides sufficient material to perform 96 reactions using the manual Precision ID Library kit (Cat. No. A26435) or 32 reactions using the automated Precision ID DL8 Library Kit (Cat. No. A33212).
Frequently asked questions (FAQs)
What are the expected library lengths with the Precision ID GlobalFiler STR NGS v2?
The expected library length ranges from ˜129-303 bp.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How many reactions can be performed with the Precision ID GlobalFiler STR NGS v2 panel?
The Precision ID GlobalFiler STR NGS v2 panel provides enough reagents for 96 reactions.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Which Ion systems can the Precision ID GlobalFiler STR NGS panel v2 be used with?
The Precision ID GlobalFiler STR NGS panel v2 is only compatible with the Ion S5 System. It is not designed for use with the Ion OneTouch2 System or Ion PGM System.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
With the Precision ID GlobalFiler STR NGS panel v2, how many samples can be multiplexed?
Up to 32 libraries can be multiplexed on an Ion 530 chip (Cat. No. A27764) with the Precision ID GlobalFiler STR NGS panel v2.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.