SYBR™ Green Fast Advanced Cells-to-CT™ Kit
Invitrogen™

SYBR™ Green Fast Advanced Cells-to-CT™ Kit

The SYBR Green Fast Advanced Cells-to-Ct Kit is a complete cell lysate system designed for gene expression analysis directly from cultured cells without RNA purification.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of ReactionsDNase
A3537940 Reactions22 μL DNase I
A35380100 Reactions55 μL DNase I
A35381400 Reactions220 μL DNase I
Catalog number A35379
Price (INR)
73,046.00
Each
No. of Reactions:
40 Reactions
DNase:
22 μL DNase I
Price (INR)
73,046.00
Each

The SYBR Green Fast Advanced Cells-to-Ct Kit is a complete cell lysate system designed for gene expression analysis directly from cultured cells without RNA purification. SYBR Green Fast Advanced Cells-to-Ct cell lysis reagents are supplied with highest sensitivity Cells-to-Ct Fast Advanced reverse transcription enzyme mix, 2X RT buffer, and best-in-class PowerUp SYBR Green Master mix. The SYBR Green Fast Advanced Cells-to-Ct Kit gives you the sensitivity you need in the fastest workflow available: get gene expressions analysis results from an entire plate of cells in about 1.5 hours.

  • Complete—optimized workflow includes cell lysis reagents with gDNA removal, new Fast Advanced RT enzyme mix, buffer, and new PowerUp SYBR Green Fast Advanced Master Mix
  • Fast—7-minute sample prep, including DNase treatment, at room temperature
  • Easy—lyse samples in a tube or directly in culture plates
  • Robust—perform gene expression analysis on 10–100,000 cells per sample; results equivalent to those from purified RNA
  • Efficient—contains sufficient reagents to generate 1000 real-time PCR results from 100 starting samples

Unique method eliminates the need to isolate RNA from cultured cells

The Cells-to-Ct method features a unique lysis reagent that lyses cells and protects RNA. DNA removal occurs simultaneously in the 5-minute lysis step to save time and improve ease of use. Lysis is then stopped by the addition of a 'Stop Solution' that irreversibly binds to the lysis reagents.

The SYBR Green Fast Advanced Cells-to-CT Kit contains Cells-to-Ct Fast Advanced reverse transcription (RT) reagents for cDNA synthesis and PowerUp SYBR Green Master Mix for real-time PCR analysis. Primer sets are provided by the user.

Simple 7-minute sample preparation

The SYBR Green Fast Advanced Cells-to-CT Kit is designed for 10–100,000 cultured cells/sample. Cells are washed in PBS and lysed in lysis solution for five minutes at room temperature; DNase treatment can be performed simultaneously. Lysis is terminated at room temperature by a 2-minute incubation with Stop Solution. The lysates are now ready for reverse transcription or storage at –20°C for a short term or -80C for a long term.

Maximize sensitivity of detection

Unlike some competitor kits that limit the amount of lysate in the RT reaction to 10%, the SYBR Green Fast Advanced Cells-to-CT Kit can accommodate 45% of the total RT reaction volume as cell lysate. Additionally, cDNA can comprise up to 30% of the real-time PCR reaction volume. The large lysate volume in the optimized RT reaction, along with the large cDNA volume in the subsequent real-time PCR using the PowerUP SYBR Green Master Mix, lead to maximum sensitivity. The master mix amplifies the target precisely and accurately, enabling the detection of small quantities of target, such as transcripts expressed at low levels.

Reduced sample loss and transfer error

Because samples can be processed directly in culture wells (96 or 384 wells), sample handling and the potential for sample loss or transfer error are minimized, facilitating rapid, high-throughput processing. Unlike old-fashioned, multi-step RNA isolation protocols, no heating, washing, or centrifugation steps are required. The kit greatly simplifies a laborious 30–60 minute process and reduces it to 7 minutes.

Proven performance, proven together

All components of the SYBR Green Fast Advanced Cells-to-CT Kit have been optimized for consistent and reliable performance. This removes the guesswork involved in assembling separate sample preparation, RT, and real-time PCR kits, giving you the sensitivity you need in the fastest sample-to-answer workflow available.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DNase22 μL DNase I
Fidelity (vs. Taq)2X
FormatTube
High-throughput CompatibilityHigh-throughput Compatible
No. of Reactions40 Reactions
Product LineCells-to-CT
Product TypeFast Advanced Cells-to-Ct Kit
Purification TargetLysate
Quantity40 reactions
Reagent TypeCell Lysis
Sample TypeMammalian Cells
Shipping ConditionDry Ice
Starting Material10-100,000 Cells
Sufficient For40 Reactions
Concentration2X
Detection MethodSYBR
For Use With (Application)Gene Expression
GC-Rich PCR PerformanceHigh
PCR Method2-step RT-qPCR
Reaction Speed7 min.
Unit SizeEach
Contents & Storage
100 μL 20X Fast Advanced RT Enzyme Mix, 2 mL 2X Fast Advanced RT Buffer, 2 mL Lysis Solution, 220 μL Stop Solution, 22 μL DNase I, 1 mL PowerUp SYBR Green Master Mix, Store at -5 to -30°C.

Frequently asked questions (FAQs)

Will the Cells-to-CT system work with my special cell line?

There is no reason why the Cells-to-CT system shouldn’t work with any cell line. However, due to differences in cell size and composition, the maximum number of cells per lysis reaction may be slightly different for different cell lines. We recommend testing for inhibition and optimal cell input by using the TaqMan Cells-to-CT and SYBR Green Cells-to-CT Control kits.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

Do the Fast Advanced Cells-to-CT Kits (Cat. No. A35374, A35379) contain RNase inhibitor?

Yes, the stop solution contains RNase inhibitor. Unfortunately, the type and concentration of the RNase inhibitor are proprietary.

Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.

Is the enzyme in the Fast Advanced Cells-to-CT kits Cat. No. A35374, A35379) sold separately?

The reverse transcription reagents can only be purchased in bulk (Cat. No. A39110).

Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.

Do the 2-step Cells-to-CT kits contain RNase inhibitor?

Yes, the Stop Solution provided in the 2-step Cells-to-CT kits contains RNase inhibitor.

Can I use the TaqMan Gene Expression Cells-to-CT kit for multiplexing?

The TaqMan Gene Expression Cells-to-CT kit has been validated for duplexing. If you want to set up a multiplex real-time PCR reaction with 3 assays, we recommend using the TaqMan Fast Advanced Cells-to-CT kit (https://www.thermofisher.com/order/catalog/product/A35374).