Qualification Services Plan for the iSort™ Automated Cell Sorter
Qualification Services Plan for the iSort™ Automated Cell Sorter
Actual product may vary
Invitrogen™
Qualification Services Plan for the iSort™ Automated Cell Sorter
Have Questions?
Catalog number A36358
Price (INR)
740,021.00
OQ/IPV, iSort
Add to cart
Price (INR)
740,021.00
OQ/IPV, iSort
Add to cart
Qualification services for the iSort Automated Cell Sorter are designed to help balance business and laboratory requirements and help you comply with global standards, country-specific regulations, and other laboratory site requirements.
Verify instrument performance and minimize downtime with IQ/OQ/IPV services
Our factory-trained and experienced engineers will conduct installation qualification (IQ), operational qualification (OQ), and instrument performance verification (IPV) on the iSort Automated Cell Sorter in your lab. These qualification services help ensure your instruments are installed, operating, and performing according to manufacturer’s specifications and help move you into production faster. Documentation for audits is provided.
When are IQ/OQ/IPV recommended?
• At installation
• After a move
When are OQ/IPV recommended?
• After planned maintenance
• After critical repairs
• After additions, changes, or upgrades to the system
• For a scheduled OQ site requirement
For more detailed information, view the figure "Qualification services for iSort Automated Cell Sorters" below and visit instrument IQ/OQ/IPV.
Certain exclusions apply. Contact your Services and Support Representative for details at compliancesales@thermofisher.com.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineiSort™
Product SizeOQ/IPV, iSort
Unit SizeOQ/IPV, iSort