Switch to CTS™ B-27™ Supplement XenoFree minus Insulin for cell therapy development and manufacturing.
B-27™ Supplement, XenoFree, minus insulin
Gibco™

B-27™ Supplement, XenoFree, minus insulin

B-27 Supplement, XenoFree, minus insulin, is a Research Use Only supplement based on the classic B-27 Supplement formulation and containsRead more
Have Questions?
Catalog NumberQuantity
A369520110 mL
Catalog number A3695201
Price (INR)
-
Quantity:
10 mL
B-27 Supplement, XenoFree, minus insulin, is a Research Use Only supplement based on the classic B-27 Supplement formulation and contains human-derived components and recombinant proteins. 

For cell therapy development and manufacturing, please see CTS B-27 Supplement, XenoFree, minus insulin.
 
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeNeural Cells
Concentration50 X
Culture TypeMammalian Cell Culture
DescriptionWithout Insulin
Product LineB-27
Quantity10 mL
Serum LevelSerum-Free
Shelf Life12 Months
Shipping ConditionDry Ice
SourceXeno-Free
Tested ForEndotoxin, Performance, Sterility, Mycoplasma
ClassificationSerum-free
FormLiquid
Product TypeCell Culture Supplement
SterilitySterile
pH6 to 8
Unit SizeEach
Contents & Storage
Store at -5°C to -20°C and protect from light.

Frequently asked questions (FAQs)

Do you offer B-27 supplements with and without insulin?

Yes, we do offer B-27 Supplement, minus insulin (Cat. Nos. A1895601 and A1895602). All our other B-27 supplements do contain insulin.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can I thaw B-27 Supplements in a 37 degrees C water bath or incubator?

We do not recommend thawing at 37 degrees C. Best results are observed upon thawing at room temperature for approximately 2 hrs or overnight at 2-8 degrees C.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How do I thaw B-27 Supplements?

We recommend thawing it at 4 degrees C overnight or at room temperature for 2 hours. We don't recommend thawing in a water bath or incubator.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can I re-freeze B-27 Supplements into smaller aliquots for future use?

Yes, we recommend that you aliquot when you first thaw it and thereafter subject the aliquot to no more than 2 additional freeze/thaw cycles.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.