ExpiSf™ Protein Production Kit
Gibco™

ExpiSf™ Protein Production Kit

Gibco™ ExpiSf™ Protein Production Kit is designed to significantly boost protein expression from high-density suspension cultures of Sf9 insect cells using the ExpiSf Expression System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A37678096 x 1000 mL (ExpiSf CD Medium), 24 x 1 mL (ExpiSf Enhancer)
A37678061 L (ExpiSf CD Medium), 4 x 1 mL (ExpiSf Enhancer)
A376780710 L (ExpiSf CD Medium), 40 mL (ExpiSf Enhancer)
A37678085 x 10 L (ExpiSf CD Medium), 5 x 40 mL (ExpiSf Enhancer)
Catalog number A3767809
Price (INR)
76,759.00
Online offer
Ends: 31-Dec-2026
113,716.00
Save 36,957.00 (32%)
Each
Quantity:
6 x 1000 mL (ExpiSf CD Medium), 24 x 1 mL (ExpiSf Enhancer)
Price (INR)
76,759.00
Online offer
Ends: 31-Dec-2026
113,716.00
Save 36,957.00 (32%)
Each

The ExpiSf Protein Production Kit is designed to significantly boost protein expression from high-density suspension cultures of Sf9 insect cells. The kit includes the chemically defined, animal origin-free ExpiSf Enhancer and ExpiSf CD Medium, which together power the highest possible level of protein expression from baculovirus-infected ExpiSf9 Cells.

The ExpiSf Protein Production Kit:

  • Delivers greater than 80% more protein yield in high-density suspension Sf9 cell culture than ExpiSf CD Medium alone
  • Includes ExpiSf Enhancer, which is specifically designed to work in concert with ExpiSf CD Medium to significantly boost protein expression from baculovirus-infected insect cell cultures
  • Demonstrates direct scalability of baculovirus infection from culture volumes of 4 mL in deep-well plates to >3 L in shake flasks

The ExpiSf Protein Production Kit is specifically designed to work with ExpiSf9 Cells in the ExpiSf Expression System. The kit supplies sufficient ExpiSf Enhancer and ExpiSf CD Medium to infect 1 L, 10 L, or 50 L of culture. ExpiSf CD Medium is also available for purchase separately.

Specifications
ClassificationAnimal Origin-Free, Chemically Defined, Protein-Free, Serum-Free
Compatible CellsInsect Cells
Expression MechanismCell-Based Expression
FormatLiquid
Product LineExpiSf
Quantity6 x 1000 mL (ExpiSf CD Medium), 24 x 1 mL (ExpiSf Enhancer)
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Expression SystemBaculovirus
Unit SizeEach
Contents & Storage
6 x 1000 mL bottle ExpiSf CD Medium, store in the dark at 2°C to 8°C; 6 units of 4 x 1 mL ExpiSf Enhancer, store (at 2°C to 8°C; Shelf Life: 12 months from date of manufacture)