SuperSignal™ West Atto Ultimate Sensitivity Substrate
Thermo Scientific™

SuperSignal™ West Atto Ultimate Sensitivity Substrate

SuperSignal West Atto Ultimate Sensitivity Chemiluminescent Substrate is an ultra-sensitive enhanced chemiluminescent (ECL) substrate for high attogram protein detection. SuperSignalRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A38556200 mL
A3855420 mL
A38555100 mL
Catalog number A38556
Price (INR)
80,535.00
Each
Quantity:
200 mL
Price (INR)
80,535.00
Each

SuperSignal West Atto Ultimate Sensitivity Chemiluminescent Substrate is an ultra-sensitive enhanced chemiluminescent (ECL) substrate for high attogram protein detection. SuperSignal West Atto substrate provides the highest-level sensitivity and better signal-to–noise ratios compared to other commercially available high-performance horseradish peroxidase (HRP) substrates.

SuperSignal West Atto Ultimate Sensitivity Chemiluminescent Substrate characteristics include:

  • Sensitivity: low-femtogram to high-attogram
  • Stability: 48 hours for working solution, one year for kit at 4°C
  • Compatibility: nitrocellulose and PVDF membranes
  • Signal duration: up to six hours
  • Recommended primary antibody concentration: 1:1,000–1:5,000 dilution (0.2–1.0 μg/mL)
  • Recommended secondary antibody concentration: 1:100,000–1:250,000 dilution (4–10 ng/mL)

Compare and learn more about our western blot chemiluminescent substrates ›

View all HRP substrates available ›

SuperSignal West Atto Ultimate Sensitivity Chemiluminescent Substrate is designed for superior sensitivity and signal-to–noise ratios for very low abundant targets or precious samples in western blot analysis. When combined with optimized antibody concentrations and blocking buffers, SuperSignal West Atto substrate enables detection of target proteins in amounts that are too small to be seen with typical high performance ECL substrates.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Kit ContentsLuminol/Enhancer, 100 mL, Stable Peroxide Buffer, 100 mL
SensitivityLow-femtogram to high-attogram
Shipping ConditionWet Ice
Sufficient For25 Mini-Gel Blots, 2000 sq. cm of Membrane
Detection MethodChemiluminescence
For Use With (Application)Western Blot
FormLiquid
Membrane CompatibilityNitrocellulose, PVDF
Product LineSuperSignal
Product TypeSubstrate
Quantity200 mL
SubstrateHorseradish Peroxidase
Unit SizeEach
Contents & Storage
SuperSignal West Atto Substrate / SuperSignal West Atto Stable Peroxide

• Substrate Solution, 100 mL
• Stable Peroxide Solution, 100 mL

Sufficient for 25 mini-gel blots; 2000 cm2 of membrane.

Store reagents at 2–8°C.

Frequently asked questions (FAQs)

How do I handle saturated or burned out bands using SuperSignal West Atto Ultimate Sensitivity Substrate?

Here are some suggestions:
- Shorten the time of exposure
- Wash blot and try using SuperSignal West Pico PLUS Chemiluminescent Substrate
- Strip the membrane using Restore Western Blot Stripping Buffer, re-probe with lesser amount of primary and secondary antibodies, and try first with SuperSignal West Pico PLUS Chemiluminescent Substrate
- Re-run the gel using lesser sample

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

How can I reduce background with SuperSignal West Atto Ultimate Sensitivity Substrate?

Here are potential causes for high background and corresponding solutions:
- Too much HRP in the system: Further dilute the HRP-conjugate, see guidelines in Table 1 in the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018858_SuperSignalWestAtto_UG.pdf).
- Inadequate blocking: Optimize blocking conditions.
- Inappropriate blocking reagent: Try a different blocking reagent. Use clear milk or StartingBlock blocking buffer instead of dry milk.
- Inadequate washing: Increase length, number or volume of washes.
- Film has been overexposed: Decrease exposure time or use Pierce Background Eliminator (Cat No. 21065).
- Concentration of antigen or antibody is too high: Decrease amount of antigen or antibody.
- Poor antibody specificity: Use SuperSignal Western Blot Enhancer (Cat. No. 46640).
We also recommend minimizing handling of the membrane, using clean gloves while handling, clean plasticware, glassware, forceps, or other contact surfaces. We also recommend trimming of the membrane to remove excess area.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Is SuperSignal West Atto Ultimate Sensitivity Substrate compatible with Restore Western Blot Stripping Buffer?

SuperSignal West Atto Ultimate Sensitivity Substrate is compatible with Restore Western Blot Stripping Buffer.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

When should I use SuperSignal West Atto Ultimate Sensitivity Substrate?

We recommend using SuperSignal West Atto Ultimate Sensitivity Substrate for detection of proteins in the following situations:
- For proteins with low concentration, not measurable by BCA or Bradford assays
- For low-abundance phosphorylated or glycosylated proteins
- For low-abundance membrane or nuclear proteins
- For low-abundance proteins being detected with a low-affinity antibody or when the antibody is precious
- When sample source is limited or when multiple conditions or assays are being tested on a single sample

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Is SuperSignal West Atto Ultimate Sensitivity Substrate affected by freeze/thaw cycles?

SuperSignal West Atto Ultimate Sensitivity Substrate can tolerate up to 3 freeze/thaw cycles without significant loss in performance.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.