Invitrogen Collibri Stranded RNA Library Prep Kit
Thermo Scientific™

Invitrogen Collibri Stranded RNA Library Prep Kit

The Invitrogen Collibri Stranded RNA Library Preparation Kit Modules are designed for a custom build of next-generation sequencing (NGS) workflow on Illumina systems. The modules originally support whole-transcriptome sequencing (human, mouse, rat) and mRNA sequencing (compatible with all species) applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeQuantity
A39118096RNA Single-indexes (Set 1-96)96 preps
A39100024Library Cleanup Kit24 preps
A39114096Library Cleanup Kit96 preps
A39115024rRNA Depletion Kit24 preps
A39115096rRNA Depletion Kit96 preps
A39117024Stranded RNA Library Prep Kit24 preps
A39117096Stranded RNA Library Prep Kit96 preps
A39118024RNA Single-indexes (Set 1-24)24 preps
A39119024RNA UD Indexes (Set 1-24)24 preps
A39119196RNA UD Indexes (Set 1-96)96 preps
Catalog number A39118096
Price (INR)
29,870.00
Online offer
Ends: 31-Mar-2026
49,783.00
Save 19,913.00 (40%)
Each
Product Type:
RNA Single-indexes (Set 1-96)
Quantity:
96 preps
Price (INR)
29,870.00
Online offer
Ends: 31-Mar-2026
49,783.00
Save 19,913.00 (40%)
Each
The Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina Systems is designed for the robust construction of cDNA libraries for strand-specific RNA sequencing on Illumina next-generation sequencing (NGS) systems. The modules bring together the superior features of SuperScript IV Reverse Transcriptase, Dynabeads magnetic particles, and Platinum SuperFi DNA Polymerase, enabling users to obtain high-quality sequencing-ready libraries.
The Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina Systems is designed for the robust construction of cDNA libraries for strand-specific RNA sequencing on Illumina next-generation sequencing (NGS) systems. The modules bring together the superior features of SuperScript IV Reverse Transcriptase, Dynabeads magnetic particles, and Platinum SuperFi DNA Polymerase, enabling users to obtain high-quality, sequencing-ready libraries.

This Collibri Stranded RNA Library Prep Kit Modules supports fast library preparation within 4.5 hours for mRNA or whole transcriptome libraries with 1–25 ng of mRNA-enriched or ribodepleted RNA sample input. Multiplexing of libraries can be carried out using up to 96 single-indexed or unique dual-indexed primers, enabling single-read or paired-end sequencing. The kit is suitable for various-quality RNA samples, including FFPE samples.

Visual cues of process integrity
For maximum convenience, visual cues of process integrity are included throughout the library preparation workflow. Monitor progress of library generation using visual feedback from dyes in critical reagents. The reaction mix changes color in response to the addition of critical components at each step to ensure that library preparation has the highest chance of success. The inert dyes do not interfere with enzymatic reactions and do not compromise sequencing results.

How it works
Full-length Illumina-compatible adaptors are introduced by PCR amplification using Platinum SuperFi DNA Polymerase to construct single-indexed or unique dual-indexed libraries compatible with single-read or paired-end sequencing. The kit contains 24 or 96 either single-indexed barcoded PCR primers (i7) pre-mixed with a universal i5 primer or unique dual-indexed PCR primers that enable multiplexing of up to 96 libraries. Optimized cleanup steps efficiently remove residual primers and adapter/primer dimers while preserving high library yields.

A related product, the Collibri Library Quantification Kit, is recommended for qPCR-based quantifications of libraries before proceeding to sequencing.

Applications
The Collibri Stranded RNA Library Prep Kit is recommended for:
• Gene expression studies
• Alternative splicing analysis
• Non-coding RNA detection and discovery
• Identification of alternative polyadenylation sites
• Gene fusion detection
• Detection of transcript isoforms

Specifications
DescriptionIndexing
For Use With (Application)RNA sequencing; whole transcriptome; mRNA sequencing; small RNA sequencing
Product LineCollibri
Product TypeRNA Single-indexes (Set 1-96)
Quantity96 preps
Sample Size96 preps
FormatKit
Unit SizeEach
Contents & Storage
Store at -20°C