KingFisher™ Flex Purification System, SmartStart, 24 deep-well head

KingFisher™ Flex Purification System, SmartStart, 24 deep-well head

Prepare samples efficiently from a variety of materials with the Thermo Scientific KingFisher Flex Purification System, which offers highly versatile,Read more
Have Questions?
Catalog NumberQuantity
A399681 package
Catalog number A39968
Price (INR)
-
Quantity:
1 package
Prepare samples efficiently from a variety of materials with the Thermo Scientific KingFisher Flex Purification System, which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein, or cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproduciblity and quality.

This package includes the KingFisher Flex Purification System and SmartStart Orientation.

Features of this KingFisher Flex Purification System package include:
• Fully automated system yields high-speed purification of nucleic acids, proteins, and cells
• High-throughput system processes up to 96 samples in less than 20 minutes
• Open and flexible system lets the customer use any magnetic particle-based kit to meet the application demands
• Easy-to-use BindIt Software provides instrument control, protocol creation, and modification
• Ready-made protocols available for different type of applications
• 1-day SmartStart Orientation to get you up and running quickly

More about the KingFisher Flex Purification System ›

Get up and running fast with SmartStart Orientation
The SmartStart Orientation included in this package is performed by a Field Application Scientist (FAS) in your lab and helps you:
• Get up and running quickly
• Get connected to your local network and Cloud
• Choose appropriate reagents and consumables to meet your needs
Content will vary depending on your level of knowledge and experience.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Collection Efficiency of Particles>95%
Depth (English)23.6 in.
Depth (Metric)60 cm
DescriptionKingFisher 24 DW plate
DisplaySTART / PAUSE / STOP / OK / TURNTABLE ROTATING CLOCKWISE / TURNTABLE ROTATING COUNTERCLOCKWISE / 4 cursor keys / LCD
For Use With (Application)Genomics and Proteomics, Target Identification, Veterinary Assays, Biomarker Discovery, Quality Control, Forensics
For Use With (Equipment)Nucleic Acid Purification Systems
Heating TemperatureAmbient +5°C to 115°C
Height (English)15 in.
Height (Metric)38 cm
Magnetic Rods24 in. one frame; interchangeable magnet heads
Particle Size (Metric)ca. >1 μm
Plate Type24 or 96-well plates, KingFisher 24 Deep Well plate (200 to 5000 μL), Microtiter 96 Deep Well plate (50 to 1000 μL), KingFisher 96 Plates (20 to 200 μL), PCR plate (20 to 100 μL)
Quantity1 package
Total Service Coverage1 year manufacturer warranty only
Weight (English)62 lb.
Weight (Metric)28 kg
Width (English)26.8 in.
Width (Metric)68 cm
Capacity24 Samples
Plates per Deck24
Volume (Metric) Processing50 to 1000 μL
Unit SizeEach
Package Components
TRN00340
Catalog Number
TRN00340
Unit Size
Qty
1