Geltrex™ Flex LDEV-Free Organoid-Qualified Reduced Growth Factor Basement Membrane Matrix
Geltrex™ Flex LDEV-Free Organoid-Qualified Reduced Growth Factor Basement Membrane Matrix
Geltrex™ Flex LDEV-Free Organoid-Qualified Reduced Growth Factor Basement Membrane Matrix
Gibco™

Geltrex™ Flex LDEV-Free Organoid-Qualified Reduced Growth Factor Basement Membrane Matrix

Geltrex Flex LDEV-Free Organoid-Qualified Reduced Growth Factor Basement Membrane Matrix is a soluble basement membrane matrix purified from EHS tumor cells used for the culture of tumoroids and organoids. Each lot is tested for the culture of mouse intestinal, liver ductal, airway, and mouse small intestinal organoids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A40000469025 mL
A40000469011 mL
A400004690310 mL
Catalog number A4000046902
Price (INR)
17,328.00
線上優惠
Ends: 31-Mar-2026
28,878.80
Save 11,550.80 (40%)
Each
Quantity:
5 mL
Price (INR)
17,328.00
線上優惠
Ends: 31-Mar-2026
28,878.80
Save 11,550.80 (40%)
Each

Gibco Geltrex Flex LDEV-Free Organoid-Qualified Reduced Growth Factor Basement Membrane Matrix is a soluble form of basement membrane that is purified from Engelbreth-Holm-Swarm (EHS) tumor cells. The major components of Geltrex Flex matrix include laminin, collagen IV, entactin, and heparan sulfate proteoglycan.

Recommended for culturing tumoroids and other organoids, organoid-qualified Geltrex Flex matrix was used to grow human lung cancer tumoroids and human colorectal cancer tumoroids in suspension and embedded culture using the OncoPro Tumoroid Culture Medium Kit. For a detailed description of OncoPro tumoroid culture, see the OncoPro Tumoroid Culture Medium Kit User Guide (Pub. No. MAN0029372). To help ensure support of organoid culture, every lot of organoid-qualified Geltrex Flex matrix is quality control–tested for stable dome formation and growth of multiple mouse organoids.

Features of Geltrex Flex LDEV-Free Organoid-Qualified Reduced Growth Factor Basement Membrane Matrix

  • Lots tested to help ensure growth of mouse intestinal, liver, ductal, and airway organoids and the expansion of mouse small intestinal organoids for more than five passages with typical organoid budding morphology
  • Each lot tested for the absence of lactose dehydrogenase elevating virus (LDEV), mycoplasma, and a broad panel of viruses and microorganisms
  • Lots tested for endotoxin, maintenance of gel stability, and HUVEC tube formation
  • Growth factor–reduced to improve lot-to-lot consistency
  • Formulated without phenol red to minimize potential for estrogen-like effects
  • Provided in recyclable glass vials
  • 1-mL, 5-mL, 10-mL sizes for maximum flexibility. Use only what you need. Reduce need for extensive aliquoting and unwanted freeze-thaws.

For the culture of primary cells, such as epithelial cells, endothelial cells, smooth muscle cells, and angiogenesis assays, neurite outgrowth assays, or bile canaliculi formation assays with polarized hepatocytes, standard Geltrex Flex matrix can be used.

For culturing hESCs or iPSCs, it is recommended to use Geltrex Flex LDEV-Free hESC-Qualified Reduced Growth Factor Basement Membrane Matrix. Each lot of hESC-qualified Geltrex Flex matrix is tested with iPSCs and H9 hESCs for at least five passages to help ensure reliability and consistency in hESC/iPSC studies.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeHuman colorectal tumoroids, human lung tumoroids, mouse intestinal organoids, mouse liver ductal organoids, mouse airway organoids, HUVEC, primary epithelial cells, endothelial cells, smooth muscle cells, hepatocytes.
Concentration9 to 13 mg/mL
Endotoxin Level≤ 4.5 EU/mL
Product TypeBasement Membrane Matrix
Purity or Quality GradeCell Culture Grade
Quantity5 mL
Shelf Life2 Years
Shipping ConditionDry Ice
SpeciesMouse
ClassificationReduced Growth Factor
Culture TypeAdherent Cell Culture, Mammalian Cell Culture, 3D Cell Culture
FormFrozen
Serum LevelSerum-free
SterilitySterile
Without AdditivesNo Phenol Red
Unit SizeEach