Efficient-Pro™ DPM Feed Enhancer
Efficient-Pro™ DPM Feed Enhancer
Gibco™
Efficient-Pro™ DPM Feed Enhancer
Efficient-Pro DPM Feed Enhancer is a dry-format next-generation Chinese Hamster Ovary (CHO) feed enhancer used in the production of monoclonal antibodies (mAbs), offering significant performance and ease-of-use improvements compared to other products.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A400017550210 L
A40001755011 L
Catalog number A4000175502
Price (INR)
485,126.00
Each
Add to cart
Quantity:
10 L
Price (INR)
485,126.00
Each
Add to cart
Designed to work optimally with Efficient-Pro Medium and Efficient-Pro Feed 3, Efficient-Pro DPM Feed Enhancer has been formulated specifically for use in CHO-K1-GS cell lines. The Efficient-Pro system increases titer and specific productivity, improves protein quality and metabolite profiles, and is easier to use and scale with convenient formats available (liquid and DPM).
  • Increase ease-of-use with DPM dry format with simplified dissolution with minimal storage
  • Sustain cell viability for longer bioreactor runs and maximized titer
  • Improve downstream outputs such as protein glycosylation, charge-variants, and aggregation
  • Speed-up process development and streamline transfer to manufacturing scale
  • Ease of use with fast hydration time to save time on workflow
Efficient-Pro DPM Feed Enhancer has been formulated as a chemically defined, animal-origin-free cell culture feed supplement used in the bioproduction of mAbs and other therapeutics in traditional batch and fed-batch bioprocessing workflows, particularly in combination with CHO-K1 cell line variants. Efficient-Pro DPM Feed Enhancer is a balanced formulation designed to meet the metabolic needs of mAb production and does not contain growth factors or insulin to support batch-to-batch consistency and production efficiency. It has been shown to improve productivity in a CHO-K1-GS cell line when paired with Efficient-Pro Feed 3 and Efficient-Pro Medium.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineCHO-K1 GS, CHO-K1
Cell TypeMammalian Cell Culture
Quantity10 L
Shipping ConditionRoom temperature
ClassificationAnimal Origin-free, Chemically-defined, Protein-free, Serum-free
FormDehydrated, powder
Product TypeCHO Media Supplement
Unit SizeEach
Contents & Storage
Store in the dark at 2°C to 8°C.