L.b. Cas12a Nuclease
Invitrogen™
L.b. Cas12a Nuclease
L.b. Cas12a Nuclease for simple, quick and specific analysis of your samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A40001796200 pmol
A400017952000 pmol
Catalog number A40001796
Price (INR)
6,154.00
Online offer
Ends: 31-Mar-2026
8,791.00
Save 2,637.00 (30%)
Each
Quantity:
200 pmol
Price (INR)
6,154.00
Online offer
Ends: 31-Mar-2026
8,791.00
Save 2,637.00 (30%)
Each
L. b. (Lachnospiraceae bacterium) Cas12a Nuclease, also known as Cpf1, is an RNA-guided endonuclease derived from the Class II type V CRISPR-Cas system. It exhibits a high degree of specificity in targeting and cleaving particular DNA sequences in genome editing. Additionally, Cas12a nuclease possesses collateral cleavage activity, resulting in the non-specific cleavage of neighboring DNA upon target recognition.
The DNA cleavage capability of Cas12a nuclease has been leveraged for various applications, including genome editing, gene regulation, and nucleic acid detection. By programming Cas12a with specific guide RNAs, targeted modifications such as gene knockout, gene insertion, or point mutations can be introduced into the target DNA sequence.
Disclaimer: The use of this product for certain applications may require a license under third party intellectual property rights and it is buyer's sole responsibility to obtain any required license for such use. Thermo Fisher Scientific makes no representation or warranty that the use of this product is free from third party intellectual property rights, and will not indemnify the buyer against any third party infringement claim arising from buyer's use of the product.
For Research Use Only.
Specifications
Product TypeStand-alone enzyme
Quantity200 pmol
Concentration10 μM
FormLiquid
Research CategoryNucleic Acid Analysis
Unit SizeEach