Expi293™ PRO Cells
Gibco™

Expi293™ PRO Cells

Expi293 PRO Cells are a next-generation HEK293 cell line for fast, high-yield protein expression. They support robust, scalable performance across a broad range of proteins and formats with improved growth kinetics and expression consistency up to 40 passages.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A400019786 x 1 mL
A400011401 mL
Catalog number A40001978
Price (INR)
271,803.00
線上優惠
Ends: 31-Mar-2026
453,004.50
Save 181,201.50 (40%)
Each
Quantity:
6 x 1 mL
Price (INR)
271,803.00
線上優惠
Ends: 31-Mar-2026
453,004.50
Save 181,201.50 (40%)
Each

Expi293 PRO Cells are derived from the HEK 293 cell line and are a core component of the Expi293 PRO Expression System. They are maintained in suspension culture and grow to high density in Expi293 PRO Expression Medium. Expi293 PRO Cells are highly transfectable and generate superior protein yields compared to standard HEK 293 cell lines in transient protein expression.

Attributes of Expi293 PRO Cells include:

  • Suspension culture cells pre-adapted to Expi293 PRO Expression Medium
  • Higher protein expression levels than other HEK 293 cells adapted to Expi293 PRO Expression Medium
  • More rapid cell growth and higher culture viabilities than standard suspension-adapted HEK 293 cells
  • Consistent transient expression performance for 40 passages beyond thaw

Frozen cells are supplied in a vial containing 1 mL of cells at 1 x 107 viable cells/mL in Expi293 PRO Expression Medium and 10% DMSO. The cells should be thawed directly into Expi293 PRO Expression Medium. The six-vial multipack eliminates the need to expand and rebank the cells in the lab.

The cells are typically grown in flasks on a shaker platform in a humidified, CO2 cell culture incubator. However, cultures are scalable from volumes of <1 mL in multiwell plates to >10 L in bioreactors. Protein harvest can be performed at 2–7 days post-transfection, depending on the protein expressed. For expression of recombinant antibodies, harvest is typically at 4–7 days post-transfection. There is no need for culture medium supplementation or batch feeding during expression.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatVials
Product LineExpi293 PRO
Product TypeHEK293 Cells
Quantity6 x 1 mL
AgeEmbryonic
Cell Line293 (HEK)
Cell TypeKidney (Embryonic)
Culture TypeSuspension Cell Culture
FormCryopreserved
GenderFemale
SpeciesHuman
TissueHuman Embryonic Kidney Tissue
Unit SizeEach
Contents & Storage
1 x 107 cells⁄mL, store in liquid nitrogen