CTS™ ACK Lysing Buffer
CTS™ ACK Lysing Buffer
Gibco™

CTS™ ACK Lysing Buffer

CTS ACK (Ammonium-Chloride-Potassium) Lysing Buffer is used for the lysis of red blood cells in samples containing white blood cells, such as EDTA-treated whole blood, buffy coats, and bone marrow.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
A4000225501100 mLBottle
A4000225502500 mLBag - BioProcess Container (BPC)
A40002255031 LBag - BioProcess Container (BPC)
A40002255042 LBag - BioProcess Container (BPC)
Catalog number A4000225501
Price (INR)
2,435.00
Online offer
Ends: 31-Mar-2026
3,478.00
Save 1,043.00 (30%)
Each
Add to cart
Quantity:
100 mL
Format:
Bottle
Price (INR)
2,435.00
Online offer
Ends: 31-Mar-2026
3,478.00
Save 1,043.00 (30%)
Each
Add to cart

Gibco Cell Therapy Systems (CTS) ACK (Ammonium-Chloride-Potassium) Lysing Buffer is used for the lysis of red blood cells in samples containing white blood cells, such as EDTA-treated whole blood, buffy coats, and bone marrow. The complete formulation and lysis protocol are available.

Closed systems compatible
CTS ACK Lysing Buffer is available in bottle and bioprocess container formats. The bioprocess container chamber is constructed from Aegis 5-14 film and includes sterile, weldable PVC/C-Flex hybrid tubing with a female Luer lock, providing versatile connection options for integrating into a closed system manufacturing process.

CTS quality
The Gibco CTS product line enables you to reduce your burden in qualifying reagents during your transition from research to clinical applications.

Products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485- and ISO 9001–certified. Gibco CTS products follow USP <1043> ancillary materials for cell, gene, and tissue-engineered products within the responsibilities applicable to a supplier.

Gibco CTS products are of high quality and are supplied with harmonized documentation such as certificates of analysis and certificates of origin.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Culture TypeMammalian cell culture
For Use With (Application)Cell and Gene Therapy Research, Development, and Manufacturing
FormatBottle
Manufacturing QualityISO13485, MDSAP, FDA-registered, 21 CFR820
Quantity100 mL
Shelf Life12 Months
Shipping ConditionAmbient
FormClear, colorless solution
Product TypeCell Culture Supplement
SterilitySterile-filtered
Sterilization Method0.2 μm Sterile-filtered
Unit SizeEach
Contents & Storage
15°C to 30°CC. Protect from light.