Expi293™ PRO Transfection Kit
Expi293™ PRO Transfection Kit
Gibco™

Expi293™ PRO Transfection Kit

The Expi293 PRO Transfection Kit enables high-efficiency delivery of DNA into HEK293 cells, supporting fast, scalable protein expression. Optimized for use with Expi293F cells, it delivers robust performance across a broad range of protein types.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSufficient For
A40002930For 50 L transfection
A40002926For 1 L transfection
A40002929For 6 L transfection
A40002927For 10 L transfection
Catalog number A40002930
Price (INR)
2,236,498.00
Online offer
Ends: 31-Mar-2026
3,727,496.00
Save 1,490,998.00 (40%)
Each
Add to cart
Sufficient For:
For 50 L transfection
Price (INR)
2,236,498.00
Online offer
Ends: 31-Mar-2026
3,727,496.00
Save 1,490,998.00 (40%)
Each
Add to cart

The Expi293 PRO Transfection Kit is a core component of the Expi293 PRO Expression System. It is designed for transient transfection of high density cultures of HEK 293 cells. The high-efficiency, cationic, lipid-based transfection reagent and transfection enhancers are designed to power the highest possible level of protein expression from Expi293 PRO cells cultured in Expi293 PRO Expression Medium.

  • Designed specifically for transfection of high-density suspension cell culture, with complimentary feed and enhancers that boost transfection performance and protein expression
  • Achieves protein yields 2- to 10-fold higher than other transfection reagents used on high-density Expi293 PRO cell cultures
  • Provides robust and reproducible transfection results, giving users greater confidence in their results
  • Scale transfections for culture volumes of less than 1 mL to greater than 10 liters, while maintaining equivalent volumetric protein yields

The Expi293 PRO Transfection Kit is specifically designed to work with Expi293 PRO cells in the Expi29 PRO Expression System. The transfection kit consists of the cationic lipid-based Expi293 PRO transfection reagent and Expi293 PRO enhancer and feed, which boost transfection, cell viability, and protein expression.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineExpi293 PRO
Product TypeTransfection Kit
PurityCell Culture Grade
QuantityFor 50 L transfection
Sufficient ForFor 50 L transfection
Cell TypeHEK293
FormatKit
PromoterpcDNA™ 3.4
Sample TypePlasmid DNA
Transfection TechniqueLipid-based Transfection, Transient Transfection
Unit SizeEach