Genexus™ Deep Clean Kit
Genexus™ Deep Clean Kit
Ion Torrent™

Genexus™ Deep Clean Kit

The Genexus™ Deep Clean Kit is a maintenance reagent kit that is used to run one deep clean on the Genexus™ Integrated Sequencer to prevent or remove templating byproduct buildup in the instrument.
Have Questions?
Catalog NumberQuantity
A400039401 Kit
Catalog number A40003940
Price (INR)
36,989.00
Each
Add to cart
Quantity:
1 Kit
Price (INR)
36,989.00
Each
Add to cart

The Genexus Deep Clean Kit is a maintenance reagent kit that is used to run one deep clean on the Genexus Integrated Sequencer to prevent or remove templating byproduct buildup in the instrument.

This deep clean can be a part of a routine maintenance clean or, if recommended by technical support, as a solution to bring the instrument back to optimal performance.

The Genexus Deep Clean Kit is designed to:

  • Minimize hands-on time (<10 minutes to run a deep clean)
  • Enable performance of the deep clean with the help of on-screen instructions
  • Reduce user error with an automated system that detects items needed for the deck

This kit is designed for use with Genexus Software 6.8.4 or later.

For Research Use Only.
Specifications
For Use With (Equipment)Genexus Integrated Sequencer
No. of Reactions1 Deep Clean Run
Quantity1 Kit
FormatKit
Sequencing TypeNext Generation Sequencing
Unit SizeEach
Contents & Storage
15°C to 30°C