Pierce™ Anti-DYKDDDDK Affinity Resin
Pierce™ Anti-DYKDDDDK Affinity Resin
Thermo Scientific™

Pierce™ Anti-DYKDDDDK Affinity Resin

Pierce Anti-DYKDDDDK Spin Columns provide a fast, convenient method for purification of DYKDDDDK-tagged proteins expressed in in vitro protein expression systems, bacteria, yeast, and mammalian cells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A400045625 Columns
A3680310 mL
A368012 mL
A36804100 mL
Catalog number A40004562
Price (INR)
189,295.00
Each
Add to cart
Quantity:
5 Columns
Price (INR)
189,295.00
Each
Add to cart

Thermo Scientific Pierce Anti-DYKDDDDK Spin Columns provide a fast, convenient method for purification of DYKDDDDK-tagged proteins expressed in in vitro protein expression systems, bacteria, yeast, and mammalian cells. The amino acid sequence DYKDDDDK, commonly known as 'FLAG', is recognized by a high-affinity rat monoclonal antibody (clone L5) that is covalently attached to UltraLink Biosupport, which is a rigid, hydrophilic, highly crosslinked, copolymeric, and porous resin with high coupling capacity.

For protein purification, resin is added to a sample containing DYKDDDDK-tagged proteins with the tag on either the N- or C-terminus. Following capture of DYKDDDDK-tagged proteins, non-specifically bound proteins can be washed away before dissociating bound target proteins with elution buffer.

Features of Pierce Anti-DYKDDDDK Affinity Resin

  • Specific—unique base beads and highly specific antibody minimize off-target binding (low non-specific binding)
  • High purity—optimized bind-wash-elute protocol enables high purity
  • High yield—special antibody conjugation method enables high yield
  • Rapid—entire purification protocol typically takes less than 40 mins
  • Economical—purification protocol allows multiple reuses
  • Flexible—available in multiple formats including bulk loose resin and spin columns

Characteristics of Pierce Anti-DYKDDDDK Affinity Resin

  • Composition: anti-DYKDDDDK antibody covalently attached to UltraLink Biosupport, a highly crosslinked rigid support
  • Particle size: 50–80 μm
  • pH stability: 1–13
  • Operating temperature: 2–30°C; do not freeze
  • Particle concentration: 50% slurry in phosphate buffered saline, 0.02% sodium azide, pH 7.5
  • Binding capacity: ≥3.0 mg DYKDDDDK-tGFP-His protein (∼32 kDa/mL settled resin)

Available formats

  • Resin slurries—2 mL, 10 mL, and 100 mL of 50% slurry in PBS with 0.02% sodium azide, pH 7.5
  • Spin columns—10 mL spin columns containing 3 mL of settled resin, supplied at 6 mL 50% slurry
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Capacity10 mL
FormLiquid Suspension
Particle Size50 to 80 μm
Product TypeAffinity Resin
Quantity5 Columns
Shipping ConditionWet Ice
Stationary PhaseAnti-DYKDDDDK monoclonal antibodies
Sufficient For≥3.0 mg DYKDDDDK-tGFP-His protein (∼32 kDa/mL settled resin)
Unit SizeEach