Pierce™ High-Capacity EDTA-Compatible Ni-IMAC Resin
Thermo Scientific™
Pierce™ High-Capacity EDTA-Compatible Ni-IMAC Resin
Pierce High-Capacity EDTA-Compatible Ni-IMAC Resin is designed for the purification of over-expressed His-tagged proteins from cell culture media.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A400045635 Columns
A5058410 mL
A5058550 mL
A50586250 mL
A50587500 mL
A400045652 Plates
Catalog number A40004563
Price (INR)
31,450.00
Each
Quantity:
5 Columns
Price (INR)
31,450.00
Each
Pierce High-Capacity EDTA-Compatible Ni-IMAC Resin is designed for the purification of over-expressed His-tagged proteins from cell culture media. The matrix is a 6% crosslinked agarose with optimal properties for batch, column-based, and research/pilot scale FPLC column purifications. The resin has high binding capacity (up to 80 mg of protein per mL of resin) and can tolerate the high EDTA and reducing agent concentrations used to inhibit metalloproteases and stabilize sensitive intracellular proteins.
Features of Pierce high-Capacity EDTA-Compatible Ni-IMAC Resin
  • Chelator stable—use up to 20 mM EDTA and DTT with no loss in performance
  • Versatile—purify both overexpressed and secreted proteins in cell culture media (e.g., Expi293, ExpiCHO, and ExpiSf expression systems)
  • High binding capacity—>80 mg/mL binding capacity for His-tagged recombinant proteins
  • Reusable—loose resin and spin column formats can be regenerated up to 10 times using the standard CIP protocol (not applicable to 96-well plates)
  • Flexible— available in multiple formats including bulk loose resin, spin columns, and 96-well spin plates
EDTA and reducing agents such as DTT and TCEP adversely affect the performance of Ni-IMAC supports by stripping off the metal. Because of its chelator stability characteristic (EDTA tolerant), Pierce High-Capacity EDTA-Compatible Ni-IMAC Resin is specially suited for purification of expressed His-tagged proteins that are secreted into cell culture media or intracellular His-tagged proteins that need the presence of EDTA to maintain stability and function. In both cases, large quantities of samples can be purified without the Ni metal stripping off of the support, thereby providing consistent high-capacity yields.
Available formats
  • Resin slurries—20 mL, 100 mL, 500 mL, and 1 L of 50% slurry in 20% ethanol
  • Spin columns—10 mL spin columns containing 1 mL of settled resin, supplied as 5 mL of 20% slurry
  • 96-well spin plates—96-well spin plates with 50 μL of settled resin, supplied as 250 μL of 20% slurry
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding PropertyChelator Stable
Matrix6% crosslinked agarose
Quantity5 Columns
Shipping ConditionWet Ice
Sufficient ForHis-tagged protein purification
Capacity (Metric)80 mg/mL
FormLiquid Suspension
Particle Size50 to 80 μm
TypeSpin Column
Unit SizeEach