Efficient-Pro™ Medium (+) Insulin
Gibco™
Efficient-Pro™ Medium (+) Insulin
Efficient-Pro Medium (+) Insulin is a next-generation Chinese Hamster Ovary (CHO) medium used in the production of proteins such as monoclonal antibodies (mAbs), offering significant performance and ease-of-use improvements compared to other products.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A400045850320 L
A40004585011000 mL
A400045850210 L
Catalog number A4000458503
Price (INR)
159,724.00
Each
Quantity:
20 L
Price (INR)
159,724.00
Each
Efficient-Pro Medium (+) Insulin is a next-generation Chinese Hamster Ovary (CHO) medium used in the production of proteins such as monoclonal antibodies (mAbs), offering significant performance and ease-of-use improvements compared to other products.
Designed to work optimally with the Efficient-Pro Feed system, Efficient-Pro Medium (+) Insulin has been formulated specifically for use with insulin-dependent CHO cell lines. The Efficient-Pro system increases titer and specific productivity, improves protein quality, and is easier to use and scale with convenient formats and multiple packaging configurations available.
Features of Efficient-Pro Medium (+) Insulin include:
  • Supports the growth and productivity of insulin-dependent CHO cell lines
  • Sustains cell viability for longer bioreactor runs and maximized titer
  • Helps speed up process development and streamline transfer to manufacturing scale
  • Increases flexibility with a range of format and packaging configurations
Efficient-Pro Medium (+) Insulin is a chemically defined and animal-origin-free (AOF) cell culture medium based on the Efficient-Pro Medium formulation with the addition of recombinant human insulin. It is designed for the bioproduction of mAbs and other therapeutic proteins in both batch and fed-batch bioprocessing workflows. It helps support the growth and productivity of insulin-dependent CHO cell lines by eliminating the need for manual insulin supplementation. This modification reduces handling steps, minimizes variability, and helps improve workflow efficiency. Designed to be paired with the Efficient-Pro Feed system, Efficient-Pro Medium (+) Insulin provides the power to help achieve high titers, specific productivity, and the simplicity to start process development faster, facilitate easier adaptation, and transfer to manufacturing scale with ease.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell LineInsulin-dependent CHO Cell Lines
Green FeaturesSustainable packaging
Product LineNext Gen CHO Media
Product TypeCHO Media
Quantity20 L
Shelf Life12 Months
Shipping ConditionRoom temperature
ClassificationChemically-defined, Animal Origin-free, Serum-free, cGMP
Culture TypeMammalian Cell Culture
FormLiquid
Serum LevelSerum-free
Unit SizeEach
Contents & Storage
Store in the dark at 2°C to 8°C.