MagMAX™ CORE ™ Disruption Modules
Applied Biosystems™
MagMAX™ CORE ™ Disruption Modules
The Applied Biosystems MagMAX CORE disruption modules are a sample preparation system designed to complement the MagMAX CORE Nucleic Acid Purification Kit.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFormatQuantity
A40004818Plate500 Reactions
A40004819Tube100 Reactions
Catalog number A40004818
Price (INR)
-
Format:
Plate
Quantity:
500 Reactions
The Applied Biosystems MagMAX CORE disruption modules are a sample preparation system designed to complement the MagMAX CORE Nucleic Acid Purification Kit. This modular technology is optimized for the purification of nucleic acids from a wide range of sample matrices commonly submitted for veterinary diagnostic testing. The MagMAX CORE disruption modules aid in the processing of hard-to-lyse organisms like Mycobacterium avium paratuberculosis (MAP).
Key features
  • Convenience—a complete solution using CORE chemistry
  • Confidence—optimized extraction of nucleic acids from difficult-to-lyse samples
  • Flexible—purchase only what you need to lyse MAP and other hard-to-lyse organisms; 100 reaction tube module for lower throughput or 500 reaction plate module for higher throughput
  • Ergonomy—streamlined sample preparation protocol versus the previous mechanical lysis module
  • Sustainability—reduced tube consumption with use of high-throughput plate module
There are two options of module depending on workflow and throughput needs:
  • MagMAX CORE Disruption Tubes Module, 100 reactions, contains bead beating tubes prefilled with zirconia beads and a 200-mL bottle of pure clarifying solution for effective disruption of individual samples
  • MagMAX CORE Disruption Plates Module, 500 reactions, contains 96-deep-well plates prefilled with zirconia beads and a 1-L bottle of pure clarifying solution for effective disruption of a higher number of samples
For Research Use Only.
Specifications
Quantity500 Reactions
ScaleMaxi
FormatPlate
Isolation TechnologyMechanical Disruption
Unit SizeEach