M. paratuberculosis qPCR Sample Input Consumables
M. paratuberculosis qPCR Sample Input Consumables
Applied Biosystems™
M. paratuberculosis qPCR Sample Input Consumables
VetMAX M. paratuberculosis qPCR Sample Input Consumables are for use with the VetMAX M. paratuberculosis 3.0 Kit.
Have Questions?
Catalog number A40004822
Price (INR)
-
Request A Quote
VetMAX M. paratuberculosis qPCR Sample Input Consumables are for use with the VetMAX M. paratuberculosis 3.0 Kit.
Included items:
  • 100 wooden tongue depressors
  • 500 50-mL conical tubes
These items help standardize the sampling process to ensure a consistent sample is entered into the MAP qPCR workflow.
  • Tongue depressors allow for easy sampling and are suitable for all textures. They are easy to scrape and no sample gets stuck in the hollow of a spoon.
  • The tubes are flat bottomed—no need for a rack during the incubation phase. Their graduations also allow for visibility of the contained volume.
The tubes accommodate enough volume to adapt to bovine, ovine, caprine, and camelidae samples.
For Research Use Only.
Specifications
Quantity500 Reactions
ScaleMidi
Unit SizeEach