CHOvantage™ GS Cell Line Development Kit
Gibco™
CHOvantage™ GS Cell Line Development Kit
The Gibco™ CHOvantage™ GS Cell Line Development Kit provides a complete, end-to-end solution for cloning and expressing recombinant proteins in Chinese hamster ovary (CHO)-derived suspension cultures. Designed for ease of use, the kit combines all the essential components for transfection, expression, clone generation, and stable cell line selection in one kit.
Have Questions?
Catalog NumberQuantity
A400060721 Kit
Catalog number A40006072
Price (INR)
4,786,859.00
Each
Quantity:
1 Kit
Price (INR)
4,786,859.00
Each
The Gibco™ CHOvantage™ GS Cell Line Development Kit provides a complete, end-to-end solution for cloning and expressing recombinant proteins in Chinese hamster ovary (CHO)-derived suspension cultures. Designed for ease of use, the kit combines all the essential components for transfection, expression, clone generation, and stable cell line selection in one kit.
Featuring cGMP CHO-K1 GS cells and an advanced transposon vector system, the kit is optimized with the Efficient-Pro™ family of media to support robust cell growth and protein production. With streamlined workflows, it enables researchers to achieve high titers while also offering simplified commercial licensing options to accelerate development from research to commercialization.
Features of the Gibco™ CHOvantage™ GS Cell Line Development Kit include:
  • Regulatory-friendly cGMP-banked CHO-K1 GS cells
  • Glutamine Synthetase (GS) selection
  • Transposon vector system
  • Efficient Pro Media and Feeds
  • Ability to achieve titers of 7+g/L in fed batch cultures
  • Proven clonal stability of up to 100 generations
  • Short development timeline; create stable pools in as little as 4 weeks
  • Access to technical support and consultation
  • Flexible, royalty-free commercial licensing
A two part Transposon Vector System for semi-targeted integration of your Gene Of Interest
The CHOvantage™ kit features a two-part transposon vector system designed for the semi-targeted integration of your gene of interest. This system includes the pCHOvantage™ DNA vector, which contains essential genetic elements such as promoters, enhancers, and GS selection markers to help ensure high expression levels of your gene of interest. Additionally, the CHOvantage™ mRNA encodes the transposase enzyme, facilitating efficient integration of the gene into the host cell genome at specific sites. This enables high transfection efficiency and more consistent clones. This innovative system helps provide reliable and efficient gene integration for your research needs.
Optimized workflow and media to streamline stable clone development
The CHOvantage™ kit offers an optimized workflow that significantly reduces development timelines, allowing you to achieve stable pools in just 10 weeks and select stable clones in as fast as 14 weeks. This efficiency translates into substantial savings on development and material costs. The workflow incorporates the Efficient Pro family of media and feeds, which are specifically tuned to increase the performance of the CHOvantage cell line. These media formulations are meticulously designed to optimize cell culture conditions and enhance productivity in biopharmaceutical manufacturing, supporting robust cell growth and high protein expression for consistent and reliable results.
Commercial licensing that is simple, flexible, and without royalties
The CHOvantage™ Kit offers a straightforward and flexible licensing approach to support your development process. Upon purchasing the kit, you receive research-use rights necessary for you or your CRO to develop a stable cell line. When transitioning to clinical or commercial stages, a simple licensing structure is available, free from royalties and multiple milestone payments. For more information on licensing, please email outlicensing@thermofisher.com.
Consistency in productivity
With the ability to achieve titers of 7+g/L in fed batch cultures, the CHOvantage™ Cell Line Development Kit provides an easy-to-use solution to help achieve your cell culture goals. Our fast and efficient workflow allows you to progress from transfection to lead clone in as little as four months. Scalable into bioreactors, the CHOvantage™ Kit consistently produces high yields over 100 generations, enabling reliable and consistent production.
Access to Gibco™ CHOvantage™ Cell Line Development Kit Technical Support
Our Field Application Scientists, with extensive experience in cell culture, process optimization, and troubleshooting, are available to support you throughout your development process. To connect with one of our Field Application Scientists, please contact your account manager or email cellculture.support@thermofisher.com for more information.
Additional information
If you need additional information, please email our support team at cellculture.support@thermofisher.com.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineCHO-K1
Cell TypeCHO
Expression SystemMammalian
For Use With (Application)Recombinant Protein Expression
Product LineCLC
Product TypeCell Line Development Kit
Quantity1 Kit
Regulatory StatusRUO
Shelf LifeVarious
SpeciesHamster
ClassificationcGMP, Chemically-defined, Serum-free
Culture TypeMammalian Cell Culture
FormCryopreserved
Serum LevelSerum-free
Unit SizeEach