MultiShot™ DH5α T1R Chemically Competent E. coli Cells, 96-well plate
MultiShot&trade; DH5&alpha; T1R Chemically Competent <i>E. coli</i> Cells, 96-well plate
Invitrogen™

MultiShot™ DH5α T1R Chemically Competent E. coli Cells, 96-well plate

MultiShot DH5α T1R Chemically Competent E. coli Cells provide reliable, high-efficiency cloning in an automation-friendly 96-well format. Ideal for high-throughput DNA manipulation and plasmid propagation using the trusted DH5α T1R strain.
Have Questions?
Catalog NumberQuantity
A400070551 x 96 Well Plate (20 μL/Well)
Catalog number A40007055
Price (INR)
52,318.00
Each
Add to cart
Quantity:
1 x 96 Well Plate (20 μL/Well)
Price (INR)
52,318.00
Each
Add to cart

Invitrogen MultiShot DH5α T1R Chemically Competent E. coli Cells are a well-known, versatile strain that is used in many everyday cloning applications.

Features of MultiShot DH5α T1R competent cells include:

  • TOP10 strain ideal for high-efficiency cloning and plasmid propagation
  • Microtiter plate format design convenient for automated high-throughput cloning
  • mcrA mutation that supports efficient transfection of methylated DNA
  • lacZΔM15 allele that permits rapid blue/white screening of transfectants
  • endA1 mutation that helps reduce endonuclease activity and improve plasmid DNA yields
  • recA1 mutation that helps reduce unwanted recombination

Invitrogen DH5α T1R competent cells are prepared by a proprietary modification of the Hanahan method. They are suitable for use in high efficiency transformations in applications where a low amount of DNA is used or to construct libraries using plasmid-derived vectors. In addition to supporting blue/white screening, recA1 and endA1 mutations in DH5α cells increase insert stability and improve the quality of plasmid DNA prepared from minipreps.

The Φ80lacZΔM15 marker provides α-complementation of the β-galactosidase gene from pUC or similar vectors and can be used for blue/white screening of colonies on bacterial plates containing Bluo-gal or X-gal. DH5α cells are capable of being transformed efficiently with large plasmids and can also serve as a host for the M13mp cloning vectors if a lawn of DH5αF'IQ (or similar) is provided to allow plaque formation.

Genotype:

F– φ80lacZΔ M15 Δ (lacZYA-argF) U169 recA1 endA1 hsdR17 (rK– mK+) phoA supE44 λ- thi–1 gyrA96 relA1

For Research Use Only.
Specifications
For Use With (Application)Cloning
IncludespUC19 DNA, S.O.C media
Product LineMultiShot
Product TypeChemically competent cells
Quantity1 x 96 Well Plate (20 μL/Well)
Shipping ConditionDry Ice
Transformation Efficiency Level≥1.0 x 108
Format96-well Plate
SpeciesE. coli
Unit SizeEach