ProFlex™ PCR System + Plastics Package, 2x384-well, EnduraPlates
Applied Biosystems™

ProFlex™ PCR System + Plastics Package, 2x384-well, EnduraPlates

This ProFlex PCR System package includes a system plus MicroAmp EnduraPlates and MicroAmp Clear Adhesive Films.Proflex PCR SystemThe ProFlex PCRRead more
Have Questions?
Catalog NumberQuantity
A411871 Package
Catalog number A41187
Price (INR)
-
Quantity:
1 Package
This ProFlex PCR System package includes a system plus MicroAmp EnduraPlates and MicroAmp Clear Adhesive Films.

Proflex PCR System
The ProFlex PCR System combines the reliability and performance you’ve come to expect from Applied Biosystems instruments with the flexible configuration and control features that adapt to your research needs:

• Interchange the 96-well 0.2-mL block with another block type if your throughput needs change
• Access the system remotely (and conveniently) through a mobile app
• Program the instrument in seconds with a simple-to-use touchscreen interface
• Simulate your old instrument with Thermal Simulation modes
• Optimize your PCR quickly and efficiently with better-than-gradient VeriFlex blocks

For detailed instrument information and specifications, see the ProFlex PCR System.

MicroAmp EnduraPlates
MicroAmp EnduraPlates offer excellent PCR or qPCR performance in formats developed to meet your experimental needs. They are a solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments. They are validated for use with Applied Biosystems PCR thermal cyclers for optimal fit and performance.

Key features:
• Optimized for performance with Applied Biosystems instruments
• Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
• Constructed to ANSI/SBS standard, well suited to high-throughput laboratories and robot handling
• Available in multiple colors for easy organization of laboratory assays
• Excellent mechanical stability and flatness for your complete workflow needs

MicroAmp Clear Adhesive Films
MicroAmp Clear Adhesive Films create a secure seal across an entire microplate to prevent evaporation and well-to-well contamination.

Key features:
• Adhesive provides tight seal over wells to prevent leaks and evaporation.
• Easy to apply and remove
• Available as Clear (economical and sticky) or Optical (ready for real-time PCR and non-tacky)

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Dimensions22 x 13 x 10.6 in. (56.5 x 33 x 27.2 cm)
Display Type8.4 in. Color TFT LCD Touchscreen
High-throughput CompatibilityHigh-throughput Compatible
Max. Ramp Rate3°C/sec. (Block), 1.6°C/sec. (Sample)
Quantity1 Package
Reaction SpeedFast, Standard
Thermal Accuracy±0.25°C (35°C to 99.9°C)
Thermal Range0°C to 100°C
Thermal Uniformity<0.5°C (20 sec. after reaching 95°C)
User InterfaceUSB, WiFi, Cloud, Ethernet
Voltage100/240 V
WarrantyLimited Warranty
Weight45 lb. (20.4 kg)
Block Configurations2 x 384-well Isothermal Blocks
Capacity768 x 20 μL tubes, 2 x 384-well plate
Product TypePCR System
Unit SizeEach
Contents & Storage
• ProFlex 2 x 384-well PCR System
• 5 x MicroAmp EnduraPlate Optical 384-well Clear Reaction Plates with Barcode (100 plates total)
• 1 x MicroAmp Clear Adhesive Films (100 films)

Frequently asked questions (FAQs)

What is the optimal ramp rate for my thermal cycler?

There is no universal ramp rate for a thermal cycler because both the block and sample ramp rates are different for each thermal cycler. The sample ramp rate changes with the volume of the sample since more time and energy are required to heat higher sample volumes. The ramp rate may also need to be changed for a particular PCR assay or primer pair if you are trying to use the same PCR assay on two different thermal cyclers, for example.

As a reference, the block ramp rate commonly used with DreamTaq Polymerase on the MiniAmp thermal cycler is 3.0 C/sec. In a 50 µL PCR reaction, this translates to a sample ramp rate of ~1 C/sec.

Please refer to the following application note for further information about thermocycler ramp rates: https://assets.thermofisher.com/TFS-Assets/LSG/brochures/Thermal_Cycler_Ramp_RatesAppNote.pdf

Find additional tips, troubleshooting help, and resources within ourPCR Thermal Cyclers Support Center.