Applied Biosystems™ RapidLINK™ Software v1.0 License
Applied Biosystems™
Applied Biosystems™ RapidLINK™ Software v1.0 License
NOTE: For use with the Applied Biosystems-branded RapidHIT ID System only (Cat. No. A41810).Applied Biosystems RapidLINK Software v1.0 is partRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A4181510 user licenses
A418131 user license
A418143 user licenses
Catalog number A41815
Price (INR)
-
Quantity:
10 user licenses
NOTE: For use with the Applied Biosystems-branded RapidHIT ID System only (Cat. No. A41810).
Applied Biosystems RapidLINK Software v1.0 is part of the Applied Biosystems RapidHIT ID System. This fast and simple system produces trusted lab-quality forensic DNA profiles in 90 minutes that are aggregated within software for direct upload to the database of choice or for further review and analysis. This data management software expedites identity confirmation securely and efficiently.
This product provides a single Applied Biosystems RapidLINK Software v1.0 serial number and a selection of quantities of user licenses.
Features of the Applied Biosystems RapidLINK Software v1.0 include:
• Securely manage one or more Applied Biosystems RapidHIT ID systems from a central location
• Review DNA profile results in real time prior to upload
• Enable or disable users in real time
• Collect and review metadata using built-in reporting tools
• Expert system-capable
• Sample/data accessioning workflow-capable
Specifications
Product TypeSoftware License
Quantity10 user licenses
Shipping ConditionRoom Temperature
Product LineRapidLINK
Software TypeLicense
Unit SizeEach
Contents & Storage
1 document letter