Product Image
Applied Biosystems™

MagMAX™ Microbiome Bead Tubes

These bead tubes are included in the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. No. A42358). They are madeRead more
Have Questions?
Catalog NumberQuantity
A42351100 tubes
Catalog number A42351
Price (INR)
16,502.00
Each
Quantity:
100 tubes
Price (INR)
16,502.00
Each
These bead tubes are included in the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. No. A42358). They are made available separately for applications that require more tubes than are provided in the kit.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput CompatibilityHigh-throughput Compatible, Manual Protocols, Automated Protocols
Product LineMagMAX
Quantity100 tubes
Shipping ConditionRoom Temperature
Sample TypeBacteria, Blood, Buccal Samples, Cells, Fungi, Liquid Samples (e.g. Serum), Viral Samples, Yeast
TargetViral DNA/RNA, Pathogen DNA/RNA, Total Nucleic Acids
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

How many beads are in each tube of the MagMAX Microbiome Bead Tubes?

The number of beads in each tube is proprietary.

Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.