MagMAX™ Viral/Pathogen Ultra Enzyme Mix
Applied Biosystems™

MagMAX™ Viral/Pathogen Ultra Enzyme Mix

This enzyme mix is included in the MagMAX Viral/Pathogen Nucleic Acid Ultra Isolation Kit.
Have Questions?
Catalog NumberQuantity
A4236650 mL
Catalog number A42366
Price (INR)
211,139.00
Online offer
Ends: 30-Sep-2026
383,888.00
Save 172,749.00 (45%)
Each
Quantity:
50 mL
Price (INR)
211,139.00
Online offer
Ends: 30-Sep-2026
383,888.00
Save 172,749.00 (45%)
Each
This enzyme mix is included in the MagMAX Viral/Pathogen Nucleic Acid Ultra Isolation Kit (Cat. No. A42356). It is made available separately for applications that require more enzyme mix than is provided in the kit.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
EfficiencyHigh
For Use With (Application)Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput CompatibilityHigh-throughput Compatible, Manual Protocols, Automated Protocols
Product LineMagMAX
Product TypeViral/Pathogen Ultra Enzyme Mix
Quantity50 mL
Shipping ConditionDry Ice
Volume (Metric)50 mL
Sample TypeLiquid Samples (e.g. Serum), Viral Samples, Buccal Samples, Blood, Cells, Bacteria, Fungi, Yeast
TargetViral DNA/RNA, Pathogen DNA/RNA, Total Nucleic Acids
Unit SizeEach
Contents & Storage
Store in freezer (-5° to -30°C).