eBioscience™ Essential Human Treg Phenotyping Panel & Staining Buffers
Invitrogen™
eBioscience™ Essential Human Treg Phenotyping Panel & Staining Buffers
The Invitrogen eBioscience Essential Human Treg Phenotyping Panel & Staining Buffers is designed for flow cytometry-based identity testing of regulatoryRead more
Have Questions?
Catalog NumberQuantity
A42924100 reactions
Catalog number A42924
Price (INR)
-
Quantity:
100 reactions
The Invitrogen eBioscience Essential Human Treg Phenotyping Panel & Staining Buffers is designed for flow cytometry-based identity testing of regulatory T-cells (Tregs). It leverages high quality eBioscience antibodies and fixation/permeabilization reagents (for the intracellular marker FOXP3) and includes a ready-to-use protocol for experimental set-up, gating strategy, and analysis.
Benefits include:
Proven performance―antibodies and supporting reagents designed for flow cytometry
Highly referenced antibodies―includes one of the most referenced clones for FOXP3
Turnkey kit and protocol―contains reagents and standardized gating recommendations needed to perform flow cytometry characterization
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
FormatKit
Quantity100 reactions
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
Product LineeBioscience
Product TypePhenotyping Panel and Staining Buffer
Unit SizeEach
Contents & Storage
  • CD4, CD25, CD127, and FOXP3 antibodies, store at 2–8°C
  • FOXP2/TF Staining Buffer set, store at 2–8°C