LV-MAX™ Lentiviral Packaging Mix

LV-MAX™ Lentiviral Packaging Mix

LV-MAX Lentiviral Packaging Mix is an optimized single formulation containing a defined mixture of three packaging plasmids: pLP1, pLP2, and pLP/VSVG.
Have Questions?
Catalog NumberQuantity
A432371.5 mL
Catalog number A43237
Price (INR)
60,003.00
Each
Quantity:
1.5 mL
Price (INR)
60,003.00
Each

The LV-MAX Lentiviral Packaging Mix is a single, optimized set of packaging plasmids, designed to streamline and enhance the production of lentiviral vectors. This mix contains a defined combination of three essential packaging plasmids: pLP1, pLP2, and pLP/VSVG, which together supply the necessary structural and replication proteins in trans when expressed in HEK293-derived Viral Production Cells. This third-generation packaging system offers enhanced biosafety, as the Rev gene is supplied on a separate plasmid, independent of other lentiviral genes, thereby significantly reducing the likelihood of recombination and producing replication-competent lentiviral particles.

Key features of the LV-MAX Lentiviral Packaging Mix

  • Optimized formulation—combines three packaging plasmids (pLP1, pLP2, and pLP/VSVG) into a single mix, saving process development time by minimizing the need to optimize plasmid ratios
  • High titer—achieves high titers of lentiviral vectors when used with the LV-MAX Lentiviral Production System

Applications

  • Gene therapy—ideal for the production of lentiviral vectors for gene therapy research and development
  • Cell therapy—supports the generation of lentiviral vectors for cell therapy applications, facilitating seamless transition from bench to clinic
  • Research and development—provides a reliable and scalable solution for viral vector production in various research settings
Specifications
Product TypeLentiviral Packaging Mix
Quantity1.5 mL
SterilityNon-sterile
Unit SizeEach
Contents & Storage
Store at -5 to -30°C. Ships on gel ice.

Frequently asked questions (FAQs)

Are the LV-MAX Lentiviral Packaging Mix (Cat. No. A43237) and ViraPower Lentiviral Packaging Mix (Cat. No. K497500) interchangeable?

Yes, they can be used interchangeably.

Find additional tips, troubleshooting help, and resources within our Transfection Support Center.

What are the major differences between LV-MAX and CTS LV- MAX products?

The LV-MAX and CTS LV- MAX products have equivalent performance. In contrast to the research use only LV-MAX products, CTS LV-MAX products have an additional intended use for cell and gene therapy applications, and are complemented with a more robust regulatory support package to enable clinical therapeutic applications. This means that to the best of our ability, we keep our CTS product specifications and release requirements up-to-date in accordance with the latest regulatory guidance specific for this intended use.

What is the intended use claim for your LV-MAX and CTS LV-MAX products?

Our LV-MAX products are intended for research use only while our CTS LV- MAX products are intended for research use or manufacture of cell, gene or tissue based products. Please refer to the individual products for further details on the intended use claim.

Citations & References (1)

Citations & References
Abstract
Effects of an indole derivative on cell proliferation, transfection, and alternative splicing in production of lentiviral vectors by transient co-transfection.
Authors:Mier NC,Roper DK
Journal:PloS one
PubMed ID:38833479
Lentiviral vectors derived from human immunodeficiency virus type I are widely used to deliver functional gene copies to mammalian cells for research and gene therapies. Post-transcriptional splicing of lentiviral vector transgene in transduced host and transfected producer cells presents barriers to widespread application of lentiviral vector-based therapies. The present study ... More