TaqMan™ MON89788 and Roundup Ready™ Soya Event ID Kit
Thermo Scientific™

TaqMan™ MON89788 and Roundup Ready™ Soya Event ID Kit

The Thermo Scientific™ TaqMan™ MON89788 and Roundup Ready™ Soya Event ID Kit enables real-time PCR detection of MON89788 and Roundup Ready™ transgenic soybean DNA in a sample. The kit detects a percentage >0.01% of MON89788 and Roundup Ready™ transgenic soybean DNA. The kit is ISO 9001 and 14001 compliant.

Have Questions?
Catalog NumberQuantity
A437201 Kit
Catalog number A43720
Price (INR)
-
Quantity:
1 Kit

The Thermo Scientific™ TaqMan™ MON89788 and Roundup Ready™ Soya Event ID Kit enables real-time PCR detection of MON89788 and Roundup Ready™ transgenic soybean DNA in a sample. The kit detects a percentage >0.01% of MON89788 and Roundup Ready™ transgenic soybean DNA. The kit is ISO 9001 and 14001 compliant.

Key features of the kit include:

  • All reagents necessary for the real‐time PCR reaction—specific FAM™-labeled probe and primers for MON89788 DNA, VIC™-labeled probe and primers for Roundup Ready™ DNA, nucleotides, MgCl2, DNA polymerase enzyme, and other buffer components.
  • An internal positive control (IPC)—NED™-labeled probe, primers, and template, to monitor for PCR inhibition.
  • A Positive Control—synthetic DNA containing target sequences corresponding to 0.1% to Roundup Ready™ soya and 0.1% MON89788. The assay is compatible with Applied Biosystems™ QuantStudio 5™ real-time PCR system.

Sample Preparation

DNA sample preparation is accomplished with the easy-to-use GMO Extraction Kit (Cat. No. 4466336). Using up to 20 g of sample rather than the smaller sample sizes accommodated by most other DNA isolation procedures minimizes sampling errors. The GMO Extraction Kit is compatible with raw and processed meat, as well as composite food samples.

Part of a Suite of Solutions For GMO Detection and Quantification

The TaqMan™ TNOS Maize Quantification Kit is just one of our many available kits for the sensitive detection of GMO in food and feed samples.

Specifications
Detection MethodPrimer-probe
No. of Reactions48
Quantity1 Kit
Sample TypePlant, food, and feed
Storage Requirements

MON89788 and RR Master Mix (store at -20°C) General Master Mix (store at -20°C upon receipt; 2-8 °C after initial use) Positive Control (store at -20°C)

Target GeneTransgenic soybean DNA
Target Specificity>0.01% of MON89788 and Roundup Ready™ transgenic soybean DNA
For Use With (Application)Food Quality Testing
Unit SizeEach