Pierce&trade; Endotoxin-Free Magnesium Chloride (MgCl<sub>2</sub>) (10 mM)
Thermo Scientific™

Pierce™ Endotoxin-Free Magnesium Chloride (MgCl2) (10 mM)

Pierce Endotoxin-Free MgCl2 is suitable for the preparation of samples for endotoxin detection (LAL) assay. As a diluent, it willRead more
Have Questions?
Catalog NumberQuantity
A438845 x 10 mL
Catalog number A43884
Price (INR)
5,243.00
Each
Quantity:
5 x 10 mL
Price (INR)
5,243.00
Each
Pierce Endotoxin-Free MgCl2 is suitable for the preparation of samples for endotoxin detection (LAL) assay. As a diluent, it will eliminate the inhibitory chelation effects of EDTA. It contains <0.005 EU/mL endotoxin.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration10 mM
DescriptionEndotoxin-Free MgCl2 buffer
For Use With (Equipment)LAL Endotoxin Assay
PackagingVial
Quantity5 x 10 mL
Unit SizeEach