RapidINTEL™ Sample Cartridge Kit
Applied Biosystems™

RapidINTEL™ Sample Cartridge Kit

Have Questions?
Catalog NumberQuantity
A4394250 cartridges
Catalog number A43942
Price (INR)
942,170.00
Each
Quantity:
50 cartridges
Price (INR)
942,170.00
Each
The Applied Biosystems RapidINTEL Sample Cartridge, 50-Pack Kit, provides 50 self-contained, easy-to-use sample cartridges that contain all necessary reagents for sample preparation, including GlobalFiler Express chemistry. With less than one minute of hands-on time, users simply insert the sample (via swab) into the sample cartridge, insert the sample cartridge into the instrument, and receive the lab-quality forensic DNA profile information within two hours.

Notes:
1. For use with the Applied Biosystems-branded RapidHIT ID System only (Cat. No. A41810).
2. Use of this sample cartridge product requires software version 1.1 or higher on your Applied Biosystems RapidHIT ID System. Contact you local support representative for more information.

Features of the kit include:
• One-minute hands-on-time workflow provided by self-contained, single-use sample cartridges
• Easy consumables information tracking and auto-selection of instrument run parameters through integrated radio frequency identification (RFID) tagging in cartridge

The kit includes:
• 50 RapidINTEL sample cartridges
• 2 positive-control ACE GlobalFiler Express sample cartridges
• 2 negative-control ACE GlobalFiler Express sample cartridges

Specifications
For Use With (Equipment)Applied Biosystems-Branded RapidHIT ID System
Product LineRapidINTEL
Product TypeSample Cartridge Kit
Quantity50 cartridges
Throughput50 Sample Cartridges
Unit SizeEach
Contents & Storage
• 50 sample cartridges
• 2 positive control cartridges
• 2 negative control cartridges

Store at 2–8°C.