Barnstead™ MegaPure™ Glass Stills
Actual product may vary
Barnstead™ MegaPure™ Glass Stills
Barnstead™ MegaPure™ Glass Stills
Thermo Scientific™

Barnstead™ MegaPure™ Glass Stills

Obtain high purity water from distillation using these glass stills, constructed of non-leaching components.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberOutput Flow RateVoltageModel
A4402670.4 gal./hr.120 VMP-1 w/6L bottle
A4401183.4 gal./hr.240 VMP-11A
A4402660.4 gal./hr.120 VMP-1
A4403670.9 gal./hr.240 VMP-3A
A4406960.9 gal./hr.208 VMP-3A
A4406971.6 gal./hr.208 VMP-6A
Catalog number A440267
Price (INR)
-
Output Flow Rate:
0.4 gal./hr.
Voltage:
120 V
Model:
MP-1 w/6L bottle
Obtain high purity water from distillation using Thermo Scientific™ Barnstead™ Mega-Pure™ Glass Stills, constructed of non-leaching components. These glass stills effectively remove inorganic solids, organics with boiling points higher than water, bacteria and pyrogens. Available in five production capacities, units are easily wall- or bench-mounted. Maximum purity is provided from the vertical condenser design. Gain easy access for cleaning with the quick release cover.
Features:
  • Contact with only glass, quartz and PTFE components ensures ultimate purity and eliminates cross-contamination.
  • Vertical condenser design provides maximum purity.
  • High temperature cutoff shuts down the still if the temperature is too high, preventing heating element burnout.
  • Quick-release cover provides easy cleaning access.
  • Connection to optional Automatic Connection System (ACS) assures a 24-hour automatic system.
  • Stills include feedwater solenoid valve for automatic operation.
  • Stills produce water with a resistivity of 1 to 10MΩ-cm, depending on feedwater quality.

MP-1:

  • Compact, easy-to-use 1L/hr. glass still
  • Available with or without a storage bottle

MP-3A, MP-6A and MP-11A:

  • For larger volume demands with storage and options (3.4 to 13L/hr.)

MP-12A:

  • Features built-in deionizer for pretreated feed to boiler
  • Distillate cooler allows product water to be used as it is produced, cooling it from 85°C (185°F) to about 55°C (131°F) for ease of handling
  • No need to wait for cooling

Ordering Information:

Optional accessories include single or dual cartridge deionizers (D440046 or D440066), adapter kits (440376 or 440375), and replacement cartridges (D400377, D400499 or D440265). Storage options include bottles (413964, 410535, 413934, 410164), or Automated Collection System (B440704). All sold separately; contact your sales representative for details.

This product is not available in all regions.

Includes:

Feed water solenoid valve for automatic operation

Requires:

Water supply and storage reservoir

Dual Solenoid required if using pretreated (DI, distilled, RO) feedwater:

  • 440236 - for use with MP-1 (240V), MP-3A, MP-6A, and MP-11A
  • RY798X2A - for use with MP-1 (120V)

Deionization Cartridges required for MP-12A:

  • D440046 - Single Cartridge Deionizer (D1)
  • D440066 - Dual Cartridge Deionizer (D2)

All accessories sold separately

Recommended for:

Buffer/reagent prep, glassware washing, dishwasher feed

Warranty: 1 year parts, 90 days labor
 

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Amperage9 A
Auto DrainNo
Depth (English)9.75 in.
Depth (Metric)24.8 cm
Electrical Requirements120 V 50/60 Hz
Frequency50/60 Hz
Height (English)34 in.
Height (Metric)86.4 cm
ModelMP-1 w/6L bottle
Voltage120 V
Width (English)18 in.
Width (Metric)45.7 cm
Flow Rate0.4 gal./hr.
Inlet Pressure20 to 100 psi
Inlet Water Temperature (Metric)4°C to 37°C
Output Flow Rate0.4 gal./hr.
TypeGlass Still
Vent Temperature85°C to 96.7°C
Unit SizeEach