QuantStudio™ 7 Pro Real-Time PCR System Extended Warranty Package, 96-well, 0.2 mL
QuantStudio™ 7 Pro Real-Time PCR System Extended Warranty Package, 96-well, 0.2 mL
QuantStudio™ 7 Pro Real-Time PCR System Extended Warranty Package, 96-well, 0.2 mL
QuantStudio™ 7 Pro Real-Time PCR System Extended Warranty Package, 96-well, 0.2 mL
Applied Biosystems™
QuantStudio™ 7 Pro Real-Time PCR System Extended Warranty Package, 96-well, 0.2 mL
The Applied Biosystems QuantStudio 7 Pro Real-Time PCR System adds innovative smart features to the real-time PCR (qPCR) workflow, allRead more
Have Questions?
Catalog NumberContent And Storage
A44557
Catalog number A44557
Price (INR)
-
The Applied Biosystems QuantStudio 7 Pro Real-Time PCR System adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint. With facial authentication, voice commands, Smart Help, multiplex capability, motorized interchangeable blocks, simplified workflow, intuitive software, automation capabilities, and large capacitive touch-screen interface, the QuantStudio 7 Pro system offers exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation. Designed for ultimate ease-of-use, the system offers high quality, high throughput, excellent reliability, and an optimal user experience for researchers who want to work smarter in the lab.
This package includes the QuantStudio 7 Pro Real-Time PCR System, SmartSmart Orientation, and 1 or 3 year extended warranty.
Features of this QuantStudio 7 Pro Real-Time PCR System package include:
• Easy and convenient hands-free operation with facial authentication and voice commands
• Reduced errors and manual entries with smart qPCR workflow and TaqMan Array Plates with RFID
• Space savings with small footprint and true standalone operation; setup, start, monitor, and analyze directly on the touchscreen
• Compatible with over 8 million TaqMan Assays for gene expression, genetic variation, gene regulation, or protein expression experiments
• 1 day SmartStart Orientation to get you up and running quickly
• 1or 3 year AB Assurance extended warranty including comprehensive service coverage and one planned maintenance visit
More information about the QuantStudio 7 Pro Real-Time PCR System ›
Get up and running fast with SmartStart Orientation
The SmartStart Orientation included in this package is performed by a Field Application Scientist (FAS) remotely or in your lab and helps you:
• Get up and running quickly
• Get connected to your local network and Cloud
• Choose appropriate reagents and consumables to meet your needs
Content will vary depending on your level of knowledge and experience.
Service plans help protect your investment
Service plans such as the AB Assurance extended warranty included in this package help to:
• Maximize system uptime with fast repair turnaround time from manufacturer-trained and certified field service engineers (FSEs)
• Reduce overall repair costs with remote instrument diagnosis and resolution
• Extend the life of your instrument and help keep it running at peak performance with annual maintenance service
During the annual maintenance, the FSE will update software if required, clean, lubricate, and align relevant parts and optical systems, functional tests as well as document tasks, results, and findings.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Customer Education1 Day at Customer Site
Display Type12 in. Color Touchscreen
For Use With (Equipment)Real Time Thermal Cyclers
QuantitySystem + SmartStart + 3 yr ext warranty
Shipping ConditionRoom Temperature
Total Service Coverage1 Year Manufacturer Warranty and 1 Year of Field Service with PM
Block FormatInterchangeable
Unit SizeEach
Package Components
QuantStudio™ 7 Pro Real-Time PCR System, 96-well, 0.2 mL
Catalog Number
A43162
Unit Size
1 system
Qty
1
TRN00303
Catalog Number
TRN00303
Unit Size
Qty
1
ZG11SCQSTUDIO7PRO
Catalog Number
ZG11SCQSTUDIO7PRO
Unit Size
Qty
3
Contents & Storage
QuantStudio 7 Pro Real Time PCR System, 96-well, 0.2 mL block with 3 year extended Warranty and SmartStart orientation
Frequently asked questions (FAQs)
What is the difference between continuous melt curves and step and hold melt curves on the Applied Biosystems Real-Time PCR Instruments?
Continuous melt curve: increases the temperature by the ramp increment (degrees C/sec) so that the temperature will change at a constant rate set in the protocol.
Step and Hold melt curve: increases the temperature by the ramp increment (degrees C) and then holds at that temperature for the time specified by the user.
The amount of data collected will depend on the filter(s) selected. The more filters you use, the longer the acquisition will take and the fewer data points will be collected.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
I recently switched the block in my real-time PCR instrument. Now it shows the status as 'not calibrated' for all the calibration plates, even though my calibrations are current. What is causing this issue?
The issue might be due to the block not being properly seated in the instrument. Please follow the steps below:
  1. Remove the block from the instrument.
  2. Reinsert the block into the instrument.
If the problem persists after reseating the block, please contact our Technical Support team for further assistance.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.