Commercial sale and application support for this item is provided by Floragenex, our global authorized distributor. Please contact Floragenex at agriseq.orders@floragenex.com for purchasing, pricing, and product inquiries.
Product Image
Ion Torrent™

Ion 530™ Chip Kit-4 Reactions for Agrigenomics

The Ion 530™ Chip Kit-4 Reactions contains 4 barcoded chips for sample tracking and sequencing of 400-600 bp fragments. TheRead more
Have Questions?
Catalog NumberQuantity
A458504 chips
Catalog number A45850
Price (INR)
-
Quantity:
4 chips
The Ion 530™ Chip Kit-4 Reactions contains 4 barcoded chips for sample tracking and sequencing of 400-600 bp fragments. The kit is intended for use with the Ion 520™ & Ion 530™ ExT Kit-Chef and the Ion S5™ and Ion S5™ XL Sequencing Systems. The Ion 530 Chip electronically detects polymerase-driven base incorporation without the use of fluorescence. By eliminating the use of an optical detection system, this advance in next-generation sequencing technology allows for rapid sequencing times of as little as 4.5 hours for 600 bp sequencing.

The Ion 530 Chip Kit-4 Reactions offers:
• Sample tracking functionality from an integrated barcode
• Read lengths of 400-600 bp to support a variety of research applications, including microbial sequencing and metagenomics
• A rapid sequencing run time of as little as 4.5 hours, generating up to 12 million reads with ExT chemistry
• Automated handling and loading of templated products for sequencing
• Compatibility with current library preparation methods

Notes:
This kit is only compatible with the Ion S5 and Ion S5 XL Systems.
Use this kit with the Ion 520 & Ion 530 ExT Kit-Chef and Torrent Suite™ Software 5.2 or higher.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Ion Proton™ System
Number of Reads per Chip15-20 M
Product TypeChip Kit-4 Reaction
Quantity4 chips
Shipping ConditionRoom Temperature
Product LineIon 530
Software TypeTorrent Suite Software 5.2
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

What is the throughput of the Ion 520 Chip?

For 200 bp sequencing, it will yield 0.6-1.0 Gb. For 400 bp sequencing, it will yield 1.2-2 Gb.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.